This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm00845399_s1
See other MELA GE Assays ›
Gene Symbol:
Mela Kcnma1
Gene Name:
melanoma antigen
potassium large conductance calcium-activated channel, subfamily M, alpha member 1
Gene Aliases:
80kDa, Ag, env, gag, gag-pol, pol
5730414M22Rik, BKCa, MaxiK, Slo, Slo1, mSlo, mSlo1
Chromosome Location:
-
Chromosome Location:
Chr.14: 23292291 - 24004951 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
-
Assay ID Mm00845399_s1
Size
Availability Inventoried
Catalog # 4331182
Price 244.00
Your Price
Online offer: 200.00
200.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Mela
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
GenBank mRNA AK090120.1 - 6452 132
AK090126.1 - 6455 132
AK090128.1 - 6456 132
BC113756.1 - 884 40047695 132
D10049.1 - 943 132
U63133.1 - 6450 132
Gene Symbol: Kcnma1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
GenBank mRNA GU441834.1 - 1341 132

Back To Top

More Information


Additional Information:

Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

Set Membership:

Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Mus musculus 1700 Inventoried

Panther Classification:

Molecular Function -

viral or transposable element protein

MicroArray Probes:

Agilent Affymetrix ABI Illumina

Array Name Probe IDs Targets Target Type
Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P142487 - -
Whole Mouse Genome G4122A A_51_P326826 D10049.1 mouse_transcriptome
Whole Mouse Genome G4122A A_51_P326826 BB795191.1 mouse_transcriptome
Whole Mouse Genome G4122A A_51_P326826 U63133.1 mouse_transcriptome
Whole Mouse Genome G4122A A_51_P326826 BB794642.1 mouse_transcriptome
Whole Mouse Genome G4122A A_51_P326826 AK090126.1 mouse_transcriptome
Array Name Probe IDs Targets Target Type
Affymetrix Mouse Gene 1.0 ST Array 10582550 - -
Affymetrix Mouse Gene 1.0 ST Array 10582547 - -
Affymetrix Mouse Gene 1.0 ST Array 10582546 - -
Affymetrix Mouse Gene 1.0 ST Array 10510223 - -
Affymetrix Mouse Gene 1.0 ST Array 10582548 - -
Affymetrix Mouse Gene 1.0 ST Array 10573866 - -
Affymetrix Mouse Gene 1.0 ST Array 10508803 - -
Affymetrix HT MG-430 PM Array Plate 1456456_PM_x_at - -
Affymetrix HT MG-430 PM Array Plate 1456182_PM_x_at - -
Affymetrix HT MG-430 PM Array Plate 1433438_PM_x_at - -
Affymetrix Mouse Gene 1.1 ST Array 10510221 - -
Affymetrix Mouse Gene 1.1 ST Array 10582549 - -
Affymetrix Mouse Gene 1.1 ST Array 10582545 - -
Mouse Genome 430 2.0 Array 1433438_PM_x_at - -
Mouse Genome 430 2.0 Array 1433438_x_at - -
Mouse Genome 430 2.0 Array 1456456_PM_x_at - -
Mouse Genome 430 2.0 Array 1456456_x_at - -
Mouse Genome 430 2.0 Array 1456182_PM_x_at - -
Mouse Genome 430 2.0 Array 1456182_x_at - -
Mouse Expression Set 430 1456456_PM_x_at - -
Mouse Expression Set 430 1433438_x_at - -
Mouse Expression Set 430 1456182_x_at - -
Mouse Expression Set 430 1456456_x_at - -
Mouse Expression Set 430 1433438_PM_x_at - -
Mouse Expression Set 430 1456182_PM_x_at - -
Murine Genome U74v2 Set 97282_at - -
Murine Genome U74v2 Set 97282_PM_at - -
Mouse Genome 430A 2.0 Array 1433438_x_at - -
Mouse Genome 430A 2.0 Array 1433438_PM_x_at - -
Mouse Genome 430A 2.0 Array 1456456_PM_x_at - -
Mouse Genome 430A 2.0 Array 1456182_PM_x_at - -
Mouse Genome 430A 2.0 Array 1456456_x_at - -
Mouse Genome 430A 2.0 Array 1456182_x_at - -
Affymetrix HT Mouse Genome 430A Array 1433438_x_at - -
Affymetrix HT Mouse Genome 430A Array 1456456_PM_x_at - -
Affymetrix HT Mouse Genome 430A Array 1456182_PM_x_at - -
Affymetrix HT Mouse Genome 430A Array 1456456_x_at - -
Affymetrix HT Mouse Genome 430A Array 1433438_PM_x_at - -
Affymetrix HT Mouse Genome 430A Array 1456182_x_at - -
Array Name Probe IDs Targets Target Type
ABI Mouse Genome Survey Microarray 641023 - -
Array Name Probe IDs Targets Target Type
MouseWG-6 v2 Expression BeadChip 104560161 - -
MouseWG-6 v2 Expression BeadChip 104850687 - -
MouseWG-6 v2 Expression BeadChip 2340438 - -
Illumina mouse-6 v1.0 expression beadchip 2340438 - -
Illumina mouse-6 v1.0 expression beadchip 104560161 - -
Illumina mouse-6 v1.0 expression beadchip 104850687 - -
Illumina mouse-6 v1.0 expression beadchip 3450717 - -
Illumina mouseRef-8 v1.0 expression beadchip 0002340438 - -
Illumina mouseRef-8 v1.0 expression beadchip 0003450717 - -
Illumina mouseRef-8 v1.1 expression beadchip ILMN_2633069 - -
Illumina mouseRef-8 v1.1 expression beadchip ILMN_1230488 - -
Illumina mouse-6 v1.1 expression beadchip ILMN_1251419 - -
Illumina mouse-6 v1.1 expression beadchip ILMN_2633069 - -
Illumina mouse-6 v1.1 expression beadchip ILMN_2485074 - -
Illumina mouse-6 v1.1 expression beadchip ILMN_1230488 - -
mouse-6 v1.1 (Illumina) GI_6715563-S - -
mouse-6 v1.1 (Illumina) scl017276.1_218-S - -
mouse-6 v1.1 (Illumina) scl0017276.1_169-S - -
mouse-6 v1.1 (Illumina) scl0017276.1_44-S - -
Illumina Sentrix mouse-6.v1 bead array 104560161 - -
Illumina Sentrix mouse-6.v1 bead array 2340438 - -
Illumina Sentrix mouse-6.v1 bead array 3450717 - -
Illumina Sentrix mouse-6.v1 bead array 104850687 - -
Illumina Sentrix MouseRef-8 Expression BeadChip scl017276.1_218-S - -
Illumina Sentrix MouseRef-8 Expression BeadChip scl0017276.1_44-S - -
Illumina Sentrix Mouse-6 Expression BeadChip (2) 104560161 - -
Illumina Sentrix Mouse-6 Expression BeadChip (2) 3450717 - -
Illumina Sentrix Mouse-6 Expression BeadChip (2) 2340438 - -
Illumina Sentrix Mouse-6 Expression BeadChip (2) 104850687 - -
Illumina MouseWG-6 v2.0 expression beadchip ILMN_1251419 - -
Illumina MouseWG-6 v2.0 expression beadchip ILMN_2485074 - -
Illumina MouseWG-6 v2.0 expression beadchip ILMN_1230488 - -
Illumina MouseWG-6 v2.0 expression beadchip ILMN_1255287 - -

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0