This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm00813703_g1
See other SLC9C1 GE Assays ›
Gene Symbol:
Slc9c1
Gene Name:
solute carrier family 9, subfamily C (Na+-transporting carboxylic acid decarboxylase), member 1
Gene Aliases:
Gm610, NHE-10, Slc9a10, sNHE, spermNHE
Chromosome Location:
Chr.16: 45535266 - 45610396 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm00813703_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Slc9c1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_198106.4 NP_932774.3 24 - 25 3327 78
GenBank mRNA AY368685.1 - 24 - 25 3287 78
BC145031.1 - 3039 78
BC145032.1 - 3150 78

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC1600000678.mm.1

Set Membership:

Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Slc9c1

Panther Classification:

Molecular Function -

transporter secondary carrier transporter

Gene Ontology Categories:

Function(s) Process(es)

transport
ion transport
cation transport
sodium ion transport
multicellular organism development
spermatogenesis
cell differentiation
sperm motility
transmembrane transport
antiporter activity
solute:proton antiporter activity

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR1600005969.mm.1 NM_198106.4.1 Refseq
Affymetrix Mouse Clariom S Array PSR1600005969.mm.1 NM_198106.4 Refseq
Affymetrix Mouse Clariom S Array PSR1600005969.mm.1 TC1600000678.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR1600005971.mm.1 NM_198106.4.1 Refseq
Affymetrix Mouse Clariom S Array PSR1600005971.mm.1 NM_198106.4 Refseq
Affymetrix Mouse Clariom S Array PSR1600005971.mm.1 TC1600000678.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR1600005972.mm.1 NM_198106.4.1 Refseq
Affymetrix Mouse Clariom S Array PSR1600005972.mm.1 NM_198106.4 Refseq
Affymetrix Mouse Clariom S Array PSR1600005972.mm.1 TC1600000678.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR1600005970.mm.1 NM_198106.4.1 Refseq
Affymetrix Mouse Clariom S Array PSR1600005970.mm.1 NM_198106.4 Refseq
Affymetrix Mouse Clariom S Array PSR1600005970.mm.1 TC1600000678.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR1600005966.mm.1 NM_198106.4.1 Refseq
Affymetrix Mouse Clariom S Array PSR1600005966.mm.1 NM_198106.4 Refseq
Affymetrix Mouse Clariom S Array PSR1600005966.mm.1 TC1600000678.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR1600005968.mm.1 NM_198106.4.1 Refseq
Affymetrix Mouse Clariom S Array PSR1600005968.mm.1 NM_198106.4 Refseq
Affymetrix Mouse Clariom S Array PSR1600005968.mm.1 TC1600000678.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR1600005967.mm.1 NM_198106.4.1 Refseq
Affymetrix Mouse Clariom S Array PSR1600005967.mm.1 NM_198106.4 Refseq
Affymetrix Mouse Clariom S Array PSR1600005967.mm.1 TC1600000678.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-649bfd9c6b-jlsz7:80/100.66.128.142:80.
git-commit: 6001d82975f0049f9efc09ec70b1b2d89229eeb3
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0