This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm00809812_s1
See other TIMM17A GE Assays ›
Gene Symbol:
Timm17a
Gene Name:
translocase of inner mitochondrial membrane 17a
Gene Aliases:
17kDa, mTim17a
Chromosome Location:
Chr.1: 135301535 - 135313737 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm00809812_s1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Timm17a
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_011590.2 NP_035720.1 6 847 94
GenBank mRNA AK003129.1 - 6 848 94
BC010830.1 - 6 835 4220707 94
BC098216.1 - 6 844 1527710 94

Back To Top

More Information


Additional Information:

Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

Set Membership:

Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Mus musculus 1700 Both primers and probe map within a single exon

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Timm17a

Panther Classification:

Molecular Function -

transporter

Gene Ontology Categories:

Function(s) Process(es)

transport
intracellular protein transport
protein transport
P-P-bond-hydrolysis-driven protein transmembrane transporter activity
enzyme binding

MicroArray Probes:

Affymetrix ABI

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0100025302.mm.1 NM_011590.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR0100025302.mm.1 NM_011590.2 Refseq
Affymetrix Mouse Clariom S Array PSR0100025302.mm.1 TC0100003125.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0100025292.mm.1 NM_011590.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR0100025292.mm.1 NM_011590.2 Refseq
Affymetrix Mouse Clariom S Array PSR0100025292.mm.1 TC0100003125.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0100025293.mm.1 NM_011590.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR0100025293.mm.1 NM_011590.2 Refseq
Affymetrix Mouse Clariom S Array PSR0100025293.mm.1 TC0100003125.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0100025296.mm.1 NM_011590.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR0100025296.mm.1 NM_011590.2 Refseq
Affymetrix Mouse Clariom S Array PSR0100025296.mm.1 TC0100003125.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0100025298.mm.1 NM_011590.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR0100025298.mm.1 NM_011590.2 Refseq
Affymetrix Mouse Clariom S Array PSR0100025298.mm.1 TC0100003125.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0100025291.mm.1 NM_011590.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR0100025291.mm.1 NM_011590.2 Refseq
Affymetrix Mouse Clariom S Array PSR0100025291.mm.1 TC0100003125.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0100025294.mm.1 NM_011590.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR0100025294.mm.1 NM_011590.2 Refseq
Affymetrix Mouse Clariom S Array PSR0100025294.mm.1 TC0100003125.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0100025295.mm.1 NM_011590.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR0100025295.mm.1 NM_011590.2 Refseq
Affymetrix Mouse Clariom S Array PSR0100025295.mm.1 TC0100003125.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0100025299.mm.1 NM_011590.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR0100025299.mm.1 NM_011590.2 Refseq
Affymetrix Mouse Clariom S Array PSR0100025299.mm.1 TC0100003125.mm.1 MTA TC
Array Name Probe IDs Targets Target Type
ABI Mouse Genome Survey Microarray 479599 - -

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-fd98957d6-rbs6c:80/100.66.128.142:80.
git-commit: 278a916a254c4ca07f9ccbb2b9efa8cd7578b5be
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0