This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm00804930_g1
See other AARS GE Assays ›
Gene Symbol:
Aars
Gene Name:
alanyl-tRNA synthetase
Gene Aliases:
AI316495, C76919, sti
Chromosome Location:
Chr.8: 111027778 - 111055569 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm00804930_g1
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Aars
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_146217.4 NP_666329.2 10 - 11 1791 79
GenBank mRNA AK036738.1 - 6 - 7 2975 79
AK044451.1 - 10 - 11 1542 79
AK085725.1 - 10 - 11 1732 79
AK150227.1 - 10 - 11 1719 79
AK158139.1 - 10 - 11 1740 79
AK192207.1 - 1 - 2 119 79
AK194276.1 - 1 - 2 119 79
AK195166.1 - 1 - 2 119 79
AK199634.1 - 1 - 2 119 79
AK202822.1 - 1 - 2 119 79
AK206120.1 - 1 - 2 119 79
AY223875.1 - 10 - 11 1731 79
BC013261.1 - 7 - 8 1016 79
BC033273.1 - 1666 4196789 79

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC0800001327.mm.1

Set Membership:

Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Aars

Panther Classification:

Molecular Function -

aminoacyl-tRNA synthetase

Gene Ontology Categories:

Function(s) Process(es)

hair follicle development
tRNA modification
translation
alanyl-tRNA aminoacylation
regulation of translational fidelity
protein folding
cerebellar Purkinje cell layer development
endoplasmic reticulum unfolded protein response
cellular response to unfolded protein
tRNA aminoacylation
response to amino acid
negative regulation of neuron apoptotic process
skin development
neuromuscular process controlling balance
neuromuscular process
tRNA binding
nucleotide binding
aminoacyl-tRNA editing activity
nucleic acid binding
RNA binding
aminoacyl-tRNA ligase activity
alanine-tRNA ligase activity
protein binding
ATP binding
amino acid binding
ligase activity
ligase activity, forming aminoacyl-tRNA and related compounds
metal ion binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0800010910.mm.1 NM_146217.4 Refseq
Affymetrix Mouse Clariom S Array PSR0800010910.mm.1 TC0800001327.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0800010908.mm.1 NM_146217.4 Refseq
Affymetrix Mouse Clariom S Array PSR0800010908.mm.1 TC0800001327.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0