This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm00804697_g1
See other ATXN3 GE Assays ›
Gene Symbol:
Atxn3
Gene Name:
ataxin 3
Gene Aliases:
2210008M02Rik, AI463012, AI647473, ATX3, MJD1, Mjd, Sca3, ataxin-3
Chromosome Location:
Chr.12: 101918901 - 101958800 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm00804697_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Atxn3
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001167914.1 NP_001161386.1 2 - 3 274 134
NM_029705.3 NP_083981.2 2 - 3 274 134
GenBank mRNA AF537188.1 - 2 - 3 188 134
AK008675.1 - 2 - 3 233 134
AK030842.1 - 2 - 3 214 134
BC049743.1 - 2 - 3 274 134
BC087880.1 - 2 - 3 269 6771498 134

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC1200002246.mm.1

Set Membership:

Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Atxn3

Panther Classification:

Molecular Function -

chromatin/chromatin-binding, or -regulatory protein

Gene Ontology Categories:

Function(s) Process(es)

microtubule cytoskeleton organization
transcription, DNA-templated
regulation of transcription, DNA-templated
proteolysis
ubiquitin-dependent protein catabolic process
misfolded or incompletely synthesized protein catabolic process
regulation of cell-substrate adhesion
actin cytoskeleton organization
cellular response to heat
monoubiquitinated protein deubiquitination
exploration behavior
proteasome-mediated ubiquitin-dependent protein catabolic process
intermediate filament cytoskeleton organization
protein K63-linked deubiquitination
protein K48-linked deubiquitination
cellular response to misfolded protein
RNA polymerase II regulatory region DNA binding
histone deacetylase activity
thiol-dependent ubiquitin-specific protease activity
protein binding
peptidase activity
cysteine-type peptidase activity
omega peptidase activity
hydrolase activity
ubiquitin protein ligase binding
identical protein binding
ATPase binding
Lys63-specific deubiquitinase activity
Lys48-specific deubiquitinase activity

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR1200015566.mm.1 NM_001167914.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR1200015566.mm.1 NM_001167914.1 Refseq
Affymetrix Mouse Clariom S Array PSR1200015566.mm.1 NM_029705.3.1 Refseq
Affymetrix Mouse Clariom S Array PSR1200015566.mm.1 NM_029705.3 Refseq
Affymetrix Mouse Clariom S Array PSR1200015566.mm.1 TC1200002246.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-649bfd9c6b-jlsz7:80/100.66.128.142:80.
git-commit: 6001d82975f0049f9efc09ec70b1b2d89229eeb3
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0