This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm00724657_m1
See other EGFLAM GE Assays ›
Gene Symbol:
Egflam
Gene Name:
EGF-like, fibronectin type III and laminin G domains
Gene Aliases:
5930412K08, AU040377
Chromosome Location:
Chr.15: 7206120 - 7398746 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm00724657_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Egflam
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001289496.1 NP_001276425.1 11 - 12 1803 68
NM_001289498.1 NP_001276427.1 11 - 12 1798 68
NM_178748.6 NP_848863.1 11 - 12 1803 68
GenBank mRNA AK031136.1 - 11 - 12 1712 68
AK033332.1 - 11 - 12 1720 68
AK037223.1 - 11 - 12 1705 68
AK041546.1 - 11 - 12 1684 68
AK087659.1 - 11 - 12 1683 68
AK158909.1 - 11 - 12 1799 68
BC095994.1 - 11 - 12 1533 6467602 68
BC150710.1 - 11 - 12 1523 9087621 68
DQ223720.1 - 11 - 12 1507 68

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC1500001171.mm.1

Set Membership:

Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Egflam

Panther Classification:

Molecular Function -

protease inhibitor

Gene Ontology Categories:

Function(s) Process(es)

positive regulation of cell-substrate adhesion
peptide cross-linking via chondroitin 4-sulfate glycosaminoglycan
extracellular matrix organization
protein binding
glycosaminoglycan binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR1500009383.mm.1 NM_001289496.1 Refseq
Affymetrix Mouse Clariom S Array PSR1500009383.mm.1 NM_001289498.1 Refseq
Affymetrix Mouse Clariom S Array PSR1500009383.mm.1 NM_178748.6 Refseq
Affymetrix Mouse Clariom S Array PSR1500009383.mm.1 TC1500001171.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0