This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm00712532_m1
See other MCM10 GE Assays ›
Gene Symbol:
Mcm10
Gene Name:
minichromosome maintenance deficient 10 (S. cerevisiae)
Gene Aliases:
2410041F14Rik, AU018508, C330019M07Rik, C79164
Chromosome Location:
Chr.2: 4990724 - 5012848 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm00712532_m1
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Mcm10
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_027290.3 NP_081566.2 17 - 18 2552 73
GenBank mRNA AK030780.1 - 17 - 18 2497 73
BC023236.1 - 9 - 10 1414 73
BC039663.1 - 3 - 4 402 73
BC064314.1 - 17 - 18 2915 73
BC094576.1 - 17 - 18 2552 6853824 73
BC120687.1 - 16 - 17 2461 40129610 73
BC120689.1 - 16 - 17 2461 40129612 73

Back To Top

More Information


Set Membership:

Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Mcm10

Gene Ontology Categories:

Function(s) Process(es)

DNA replication
DNA replication initiation
cell proliferation
DNA binding
DNA replication origin binding
double-stranded DNA binding
single-stranded DNA binding
identical protein binding
metal ion binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0