This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm00671657_m1
See other MPRIP GE Assays ›
Gene Symbol:
Mprip
Gene Name:
myosin phosphatase Rho interacting protein
Gene Aliases:
9530046C02, AA536749, AI647711, C76423, RIP3, Rhoip3, mKIAA0864, p116Rip
Chromosome Location:
Chr.11: 59662473 - 59780860 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm00671657_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Mprip
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_012027.2 NP_036157.2 9 - 10 1482 65
NM_201245.3 NP_957697.1 9 - 10 1482 65
GenBank mRNA AK035435.1 - 3 - 4 772 65
AK154807.1 - 10 - 11 1489 65
AK155630.1 - 9 - 10 1263 65
AK158296.1 - 9 - 10 1471 65
AK162530.1 - 6 - 7 813 65
AK170849.1 - 9 - 10 1466 65
AK171089.1 - 9 - 10 1471 65
BC039932.1 - 8 - 9 1006 65
BC049803.1 - 9 - 10 1008 65
BC063067.1 - 9 - 10 1445 5721396 65
U73200.1 - 9 - 10 1342 65

Back To Top

More Information


Set Membership:

Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Mprip

Panther Classification:

Molecular Function -

actin or actin-binding cytoskeletal protein

Gene Ontology Categories:

Function(s) Process(es)

positive regulation of protein phosphorylation
maintenance of protein location in cell
negative regulation of myosin-light-chain-phosphatase activity
actin binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0