This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm00660264_g1
See other HMBS GE Assays ›
Gene Symbol:
Hmbs
Gene Name:
hydroxymethylbilane synthase
Gene Aliases:
PBGD, T25658, Ups, Uros1
Chromosome Location:
Chr.9: 44336348 - 44344228 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm00660264_g1
Size
Availability Made To Order
Catalog # 4351372
Price 4,408.00
Your Price
Online offer:
4408.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Hmbs
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001110251.1 NP_001103721.1 5 - 6 393 64
NM_013551.2 NP_038579.2 5 - 6 451 64
GenBank mRNA AK143558.1 - 5 - 6 318 64
AK151020.1 - 5 - 6 417 64
AK153884.1 - 5 - 6 383 64
AK166734.1 - 5 - 6 349 64
AK167702.1 - 5 - 6 410 64
AK184386.1 - 2 - 3 166 64
BC003861.1 - 5 - 6 411 3487642 64

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC0900002158.mm.1

Set Membership:

Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Hmbs

Panther Classification:

Molecular Function -

deaminase

Gene Ontology Categories:

Function(s) Process(es)

porphyrin-containing compound biosynthetic process
heme biosynthetic process
peptidyl-pyrromethane cofactor linkage
tetrapyrrole biosynthetic process
hydroxymethylbilane synthase activity
uroporphyrinogen-III synthase activity
transferase activity
carboxylic acid binding
amine binding
coenzyme binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0900017929.mm.1 NM_001110251.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0900017929.mm.1 NM_001110251.1 Refseq
Affymetrix Mouse Clariom S Array PSR0900017929.mm.1 NM_013551.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR0900017929.mm.1 NM_013551.2 Refseq
Affymetrix Mouse Clariom S Array PSR0900017929.mm.1 TC0900002158.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-649bfd9c6b-jlsz7:80/100.66.128.142:80.
git-commit: 6001d82975f0049f9efc09ec70b1b2d89229eeb3
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0