This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm00624780_g1
See other STAC3 GE Assays ›
Gene Symbol:
Stac3
Gene Name:
SH3 and cysteine rich domain 3
Gene Aliases:
9830125E18
Chromosome Location:
Chr.10: 127501617 - 127508823 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm00624780_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Stac3
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_177707.3 NP_808375.1 7 - 8 1029 57
GenBank mRNA AK036516.1 - 7 - 8 1030 57
BC067208.1 - 7 - 8 809 30286602 57

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC1000001565.mm.1

Set Membership:

Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Stac3

Gene Ontology Categories:

Function(s) Process(es)

skeletal muscle contraction
neuromuscular synaptic transmission
intracellular signal transduction
skeletal muscle fiber development
identical protein binding
metal ion binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR1000011215.mm.1 NM_177707.3.1 Refseq
Affymetrix Mouse Clariom S Array PSR1000011215.mm.1 NM_177707.3 Refseq
Affymetrix Mouse Clariom S Array PSR1000011215.mm.1 TC1000001565.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR1000011214.mm.1 NM_177707.3.1 Refseq
Affymetrix Mouse Clariom S Array PSR1000011214.mm.1 NM_177707.3 Refseq
Affymetrix Mouse Clariom S Array PSR1000011214.mm.1 TC1000001565.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-649bfd9c6b-jlsz7:80/100.66.128.142:80.
git-commit: 6001d82975f0049f9efc09ec70b1b2d89229eeb3
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0