This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm00624727_s1
See other ATMIN GE Assays ›
Gene Symbol:
Atmin
Gene Name:
ATM interactor
Gene Aliases:
Asciz, gpg6, mKIAA0431
Chromosome Location:
Chr.8: 116943386 - 116960445 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm00624727_s1
Size
Availability Inventoried
Catalog # 4331182
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Atmin
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_177700.4 NP_808368.3 4 2546 70
GenBank mRNA AK087334.1 - 1 1857 70
AK122281.1 - 4 2428 70
AX775769.1 - 4 2546 70
BC025922.1 - 1 1152 70
BC058974.1 - 4 1906 70
BC060631.1 - 4 2531 6847850 70

Back To Top

More Information


Additional Information:

Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

Set Membership:

Ambion siRNA available Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Detect Genomic DNA Mus musculus 1700 Inventoried Both primers and probe map within a single exon

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Atmin

Gene Ontology Categories:

Function(s) Process(es)

transcription, DNA-templated
regulation of transcription, DNA-templated
cellular response to DNA damage stimulus
positive regulation of transcription, DNA-templated
cilium morphogenesis
transcription regulatory region DNA binding
dynein binding
metal ion binding

MicroArray Probes:

Agilent Affymetrix ABI Illumina

Array Name Probe IDs Targets Target Type
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P26199827 - -
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P26199824 - -
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P26199828 - -
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P26199826 - -
Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2043434 BC060631.1 mouse_transcriptome
Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2043434 BC058974.1 mouse_transcriptome
Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2043434 AK122281.1 mouse_transcriptome
Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2043434 NM_177700.4 mouse_transcriptome
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 886259 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 425473 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 854896 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 41381 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 971339 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 393648 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 971660 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 487927 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 652374 - -
Agilent SurePrint G3 Mouse CGH Microarray 4x180K 808 - -
Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P111273 - -
Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P156346 - -
SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2043434 BC060631.1 mouse_transcriptome
SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2043434 BC058974.1 mouse_transcriptome
SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2043434 AK122281.1 mouse_transcriptome
SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2043434 NM_177700.4 mouse_transcriptome
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26199838 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26199824 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26199831 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26199835 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26199842 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26199827 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26199836 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26199833 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26199821 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26199829 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26199823 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26199840 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26199820 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26199828 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26199816 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26199819 - -
Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P06655506 - -
Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P156346 - -
Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P111273 - -
Whole Mouse Genome G4122A A_51_P339824 BC060631.1 mouse_transcriptome
Whole Mouse Genome G4122A A_51_P339824 BC058974.1 mouse_transcriptome
Whole Mouse Genome G4122A A_51_P339824 AK087334.1 mouse_transcriptome
Whole Mouse Genome G4122A A_51_P339824 AK122281.1 mouse_transcriptome
Whole Mouse Genome G4122A A_51_P339824 NM_177700.4 mouse_transcriptome
Whole Mouse Genome G4122A A_52_P342773 BC060631.1 mouse_transcriptome
Whole Mouse Genome G4122A A_52_P342773 BC058974.1 mouse_transcriptome
Whole Mouse Genome G4122A A_52_P342773 AK122281.1 mouse_transcriptome
Whole Mouse Genome G4122A A_52_P342773 NM_177700.4 mouse_transcriptome
Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P113295 - -
Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P156346 - -
Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P111273 - -
Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P06974823 - -
Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P07404199 - -
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P26199824 - -
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P26199828 - -
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P26199827 - -
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P26199826 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26199836 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26199949 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26199895 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26199969 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26199847 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26199942 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26199963 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26199938 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26199868 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26199859 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26199817 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26199824 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26199831 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26199908 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26199934 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26199864 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26199929 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26199954 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26199873 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26199915 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26199953 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26199983 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26199981 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26199843 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26199941 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26199919 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26199833 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26199917 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26199874 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26199936 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26199960 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26199913 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26199829 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26199945 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26199900 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26199861 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26199924 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26199857 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26199978 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26199902 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26199987 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26199985 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26199894 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26199889 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26199816 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26199925 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26199958 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26199970 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26199827 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26199939 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26199878 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26199927 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26199835 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26199849 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26199876 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26199885 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26199897 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26199884 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26199845 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26199881 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26199904 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26199932 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26199867 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26199922 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26199901 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26199906 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26199982 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26199879 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26199984 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26199852 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26199911 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P26199838 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P07444896 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P07401794 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P26199899 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P26199835 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P26199863 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P26199829 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P26199843 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P26199866 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P07404198 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P26199902 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P26199869 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P26199892 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P26199894 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P26199850 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P26199819 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P26199849 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P26199837 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P07368221 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P26199864 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P07315121 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P07426645 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P26199821 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P26199868 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P26199845 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P26199898 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P26199854 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P26199831 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P26199906 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P26199852 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P26199862 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P26199861 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P26199901 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P26199874 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P26199897 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P26199870 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P26199873 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P07444894 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P26199859 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P07201014 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P26199833 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P26199820 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P26199827 - -
Array Name Probe IDs Targets Target Type
Affymetrix Mouse Gene 1.0 ST Array 10575748 - -
Affymetrix Mouse Gene 1.0 ST Array 10575746 - -
Affymetrix Mouse Gene 1.0 ST Array 10575747 - -
Affymetrix Mouse Gene 1.0 ST Array 10575749 - -
Affymetrix HT MG-430 PM Array Plate 1434381_PM_at - -
Affymetrix HT MG-430 PM Array Plate 1457629_PM_at - -
Affymetrix HT MG-430 PM Array Plate 1434143_PM_at - -
Affymetrix Mouse Gene 1.1 ST Array 10575745 - -
Mouse Genome 430 2.0 Array 1457629_PM_at - -
Mouse Genome 430 2.0 Array 1457629_at - -
Mouse Genome 430 2.0 Array 1434143_at - -
Mouse Genome 430 2.0 Array 1434143_PM_at - -
Mouse Genome 430 2.0 Array 1434381_PM_at - -
Mouse Genome 430 2.0 Array 1434381_at - -
Affymetrix GeneChip Mouse Genome 430 2.0 Array 234776_at - -
Affymetrix GeneChip Mouse Genome 430 2.0 Array 234776_PM_at - -
Mouse Expression Set 430 1434143_PM_at - -
Mouse Expression Set 430 1434143_at - -
Mouse Expression Set 430 1457629_PM_at - -
Mouse Expression Set 430 1434381_at - -
Mouse Expression Set 430 1434381_PM_at - -
Mouse Expression Set 430 1457629_at - -
Murine Genome U74v2 Set 130730_f_at - -
Murine Genome U74v2 Set 167387_at - -
Murine Genome U74v2 Set 130730_PM_f_at - -
Murine Genome U74v2 Set 167387_PM_at - -
Mouse Genome 430A 2.0 Array 1434143_at - -
Mouse Genome 430A 2.0 Array 1434143_PM_at - -
Affymetrix HT Mouse Genome 430A Array 1434143_PM_at - -
Affymetrix HT Mouse Genome 430A Array 1434143_at - -
Affymetrix HT Mouse Genome 430B Array 1434381_at - -
Affymetrix HT Mouse Genome 430B Array 1434381_PM_at - -
Affymetrix HT Mouse Genome 430B Array 1457629_PM_at - -
Affymetrix HT Mouse Genome 430B Array 1457629_at - -
Array Name Probe IDs Targets Target Type
ABI Mouse Genome Survey Microarray 846810 - -
Array Name Probe IDs Targets Target Type
MouseWG-6 v2 Expression BeadChip 1690121 - -
Illumina mouse-6 v1.0 expression beadchip 1690121 - -
Illumina mouseRef-8 v1.0 expression beadchip 0001690121 - -
Illumina mouseRef-8 v1.1 expression beadchip ILMN_2701824 - -
Illumina mouse-6 v1.1 expression beadchip ILMN_2701824 - -
mouse-6 v1.1 (Illumina) scl33277.4_326-S - -
Illumina Sentrix mouse-6.v1 bead array 1690121 - -
Illumina Sentrix MouseRef-8 Expression BeadChip scl33277.4_326-S - -
Illumina Sentrix Mouse-6 Expression BeadChip (2) 1690121 - -
MouseRef-8 v2 Expression BeadChip ILMN_3146362 - -
MouseRef-8 v2 Expression BeadChip ILMN_3068179 - -
MouseRef-8 v2 Expression BeadChip ILMN_2701824 - -
Illumina MouseWG-6 v2.0 expression beadchip ILMN_3068179 - -
Illumina MouseWG-6 v2.0 expression beadchip ILMN_3146362 - -
Illumina MouseWG-6 v2.0 expression beadchip ILMN_2701824 - -
Illumina Sentrix Mouse-6 Expression BeadChip (3) scl33277.4_326-S - -

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-7db4fb6694-gx648:80/100.66.128.142:80.
git-commit: 672a5a8c50d6b616e949be631a941c86748ade1b
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/search_redesign_header