This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm00622843_s1
See other 9530091C08RIK GE Assays ›
Gene Symbol:
9530091C08Rik
Gene Name:
RIKEN cDNA 9530091C08 gene
Gene Aliases:
-
Chromosome Location:
Chr.9: 68765346 - 68773322 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm00622843_s1
Best Coverage
Size
Availability Inventoried
Catalog # 4331182
Price 244.00
Your Price
Online offer: 200.00
200.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: 9530091C08Rik
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NR_033299.1 1 2753 92
GenBank mRNA AK036703.1 - 1 92
AK046458.1 - 1 2752 92

Back To Top

More Information


Additional Information:

Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

Set Membership:

Amplicon length between 86 and 100 Detect Genomic DNA Recommended Assay Mus musculus 1700 Inventoried Both primers and probe map within a single exon

MicroArray Probes:

Agilent Affymetrix ABI Illumina

Array Name Probe IDs Targets Target Type
SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P517011 NR_033299.1 mouse_transcriptome
SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P517011 AK036703.1 mouse_transcriptome
SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P517011 AK046458.1 mouse_transcriptome
SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P517011 NR_033299.1 mouse_transcriptome
SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P517011 AK036703.1 mouse_transcriptome
SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P517011 AK046458.1 mouse_transcriptome
Whole Mouse Genome G4122A A_52_P517011 NR_033299.1 mouse_transcriptome
Whole Mouse Genome G4122A A_52_P517011 AK036703.1 mouse_transcriptome
Whole Mouse Genome G4122A A_52_P517011 AK046458.1 mouse_transcriptome
Whole Mouse Genome G4122A A_52_P517011 NR_033299.1 mouse_transcriptome
Whole Mouse Genome G4122A A_52_P517011 AK036703.1 mouse_transcriptome
Whole Mouse Genome G4122A A_52_P517011 AK046458.1 mouse_transcriptome
Whole Mouse Genome G4122A A_52_P191210 NR_033299.1 mouse_transcriptome
Whole Mouse Genome G4122A A_52_P191210 AK028720.1 mouse_transcriptome
Whole Mouse Genome G4122A A_52_P434499 NR_033299.1 mouse_transcriptome
Whole Mouse Genome G4122A A_52_P434499 AK046458.1 mouse_transcriptome
Array Name Probe IDs Targets Target Type
Affymetrix Mouse Gene 1.1 ST Array 10586718 - -
Affymetrix Mouse Clariom S Array PSR0900006987.mm.1 NR_033299.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0900006987.mm.1 NR_033299.1 Refseq
Affymetrix Mouse Clariom S Array PSR0900006987.mm.1 TC0900000895.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0900006988.mm.1 NR_033299.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0900006988.mm.1 NR_033299.1 Refseq
Affymetrix Mouse Clariom S Array PSR0900006988.mm.1 TC0900000895.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0900006989.mm.1 NR_033299.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0900006989.mm.1 NR_033299.1 Refseq
Affymetrix Mouse Clariom S Array PSR0900006989.mm.1 TC0900000895.mm.1 MTA TC
Array Name Probe IDs Targets Target Type
ABI Mouse Genome Survey Microarray 348000 - -
Array Name Probe IDs Targets Target Type
MouseWG-6 v2 Expression BeadChip 3840373 - -
MouseWG-6 v2 Expression BeadChip 106200528 - -
MouseWG-6 v2 Expression BeadChip 101400687 - -
Illumina mouseRef-8 v1.1 expression beadchip ILMN_2699803 - -
Illumina mouse-6 v1.1 expression beadchip ILMN_2699803 - -
Illumina mouse-6 v1.1 expression beadchip ILMN_2437257 - -
MouseRef-8 v2 Expression BeadChip ILMN_2699803 - -

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-69fc894774-tvlwx:80/100.66.128.142:80.
git-commit: 8e82defa9bf25b3b77800b5e306013d4ba941391
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0