This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm00620921_s1
See other RAPGEF4OS3 GE Assays ›
Gene Symbol:
Rapgef4os3
Gene Name:
Rap guanine nucleotide exchange factor (GEF) 4, opposite strand 3
Gene Aliases:
4732447D17Rik
Chromosome Location:
-
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm00620921_s1
Best Coverage
Size
Availability Inventoried
Catalog # 4331182
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Rapgef4os3
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
GenBank mRNA AK076344.1 - 2 3101 84

Back To Top

More Information


Set Membership:

Amplicon length between 71 and 85 3' Most Detect Genomic DNA Recommended Assay Mus musculus 1700 Inventoried Both primers and probe map within a single exon

MicroArray Probes:

ABI Illumina

Array Name Probe IDs Targets Target Type
ABI Mouse Genome Survey Microarray 433508 - -
ABI Mouse Genome Survey Microarray 766276 - -
Array Name Probe IDs Targets Target Type
MouseWG-6 v2 Expression BeadChip 100940452 - -

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-69fc894774-tvlwx:80/100.66.128.142:80.
git-commit: 8e82defa9bf25b3b77800b5e306013d4ba941391
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0