This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm00617824_m1
See other ZDHHC2 GE Assays ›
Gene Symbol:
Zdhhc2
Gene Name:
zinc finger, DHHC domain containing 2
Gene Aliases:
5730415P04Rik, 6430583A19Rik
Chromosome Location:
Chr.8: 40423000 - 40489592 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm00617824_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Zdhhc2
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_178395.3 NP_848482.1 3 - 4 483 133
GenBank mRNA AK018284.1 - 455 133
AK030662.1 - 3 - 4 484 133
AK044652.1 - 3 - 4 477 133
AK046533.1 - 3 - 4 482 133
AK136296.1 - 3 - 4 482 133
BC117761.1 - 4 - 5 245 133
BC127156.1 - 4 - 5 317 40090859 133

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC0800003224.mm.1

Set Membership:

Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Zdhhc2

Gene Ontology Categories:

Function(s) Process(es)

protein palmitoylation
zinc ion binding
palmitoyltransferase activity
transferase activity
transferase activity, transferring acyl groups
protein-cysteine S-palmitoyltransferase activity
metal ion binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0800003260.mm.1 NM_178395.3.1 Refseq
Affymetrix Mouse Clariom S Array PSR0800003260.mm.1 NM_178395.3 Refseq
Affymetrix Mouse Clariom S Array PSR0800003260.mm.1 TC0800003224.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-69fc894774-tvlwx:80/100.66.128.142:80.
git-commit: 8e82defa9bf25b3b77800b5e306013d4ba941391
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0