This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm00614092_g1
See other SIX3OS1 GE Assays ›
Gene Symbol:
Six3os1
Gene Name:
SIX homeobox 3, opposite strand 1
Gene Aliases:
D17Mgi26, E130112H22Rik, Rncr1, Six3os
Chromosome Location:
Chr.17: 85601932 - 85618396 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm00614092_g1
Size
Availability Made To Order
Catalog # 4426961
Price 4,437.00
Your Price
Online offer:
4437.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Six3os1
Interrogated Sequence Translated Protein RNADB ID Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NR_015387.2
1 - 2 342 69
GenBank mRNA AK044330.1 - LIT3371
RNCR1
1 - 2 312 69
AY589791.1 -
1 - 2 311 69
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
GenBankmRNA BB642261.1 - 1 - 2 312 Chr. 17 - 85617270 - 85618365 [-] on Build GRCm38 69
GenBankmRNA BE954756.1 - 1 - 2 221 Chr. 17 - 85617552 - 85618275 [-] on Build GRCm38 69
GenBankmRNA BE335872.1 - 1 - 2 222 Chr. 17 - 85617367 - 85618276 [-] on Build GRCm38 69

Adjacent Gene Details

Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
Upstream calmodulin-lysine N-methyltransferase Camkmt 1700106N22Rik,
AI480743,
AV099781,
CLNMT,
CaM KMT
-158.7kb Anti-sense Chr. 17 - 85090592 - 85458589 on NCBI Build GRCm38
Downstream sine oculis-related homeobox 3 Six3 E130112M24Rik,
Six3a,
Six3alpha,
Six3b,
Six3beta
2.5kb Anti-sense Chr. 17 - 85620834 - 85626191 on NCBI Build GRCm38

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC1700002661.mm.1

Set Membership:

Amplicon length less than or equal to 70 Detect Genomic DNA Mus musculus Probe spans exons

Gene Ontology Categories:

Function(s) Process(es)

regulation of photoreceptor cell differentiation
protein binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR1700024218.mm.1 NR_015385.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR1700024218.mm.1 NR_015385.2 Refseq
Affymetrix Mouse Clariom S Array PSR1700024218.mm.1 NR_015386.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR1700024218.mm.1 NR_015386.2 Refseq
Affymetrix Mouse Clariom S Array PSR1700024218.mm.1 NR_015387.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR1700024218.mm.1 NR_015387.2 Refseq
Affymetrix Mouse Clariom S Array PSR1700024218.mm.1 NR_038082.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR1700024218.mm.1 NR_038082.1 Refseq
Affymetrix Mouse Clariom S Array PSR1700024218.mm.1 NR_038083.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR1700024218.mm.1 NR_038083.1 Refseq
Affymetrix Mouse Clariom S Array PSR1700024218.mm.1 NR_038084.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR1700024218.mm.1 NR_038084.1 Refseq
Affymetrix Mouse Clariom S Array PSR1700024218.mm.1 NR_038085.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR1700024218.mm.1 NR_038085.1 Refseq
Affymetrix Mouse Clariom S Array PSR1700024218.mm.1 NR_038086.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR1700024218.mm.1 NR_038086.1 Refseq
Affymetrix Mouse Clariom S Array PSR1700024218.mm.1 TC1700002661.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR1700024220.mm.1 NR_015385.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR1700024220.mm.1 NR_015385.2 Refseq
Affymetrix Mouse Clariom S Array PSR1700024220.mm.1 NR_015386.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR1700024220.mm.1 NR_015386.2 Refseq
Affymetrix Mouse Clariom S Array PSR1700024220.mm.1 NR_015387.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR1700024220.mm.1 NR_015387.2 Refseq
Affymetrix Mouse Clariom S Array PSR1700024220.mm.1 NR_038082.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR1700024220.mm.1 NR_038082.1 Refseq
Affymetrix Mouse Clariom S Array PSR1700024220.mm.1 NR_038083.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR1700024220.mm.1 NR_038083.1 Refseq
Affymetrix Mouse Clariom S Array PSR1700024220.mm.1 NR_038084.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR1700024220.mm.1 NR_038084.1 Refseq
Affymetrix Mouse Clariom S Array PSR1700024220.mm.1 NR_038085.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR1700024220.mm.1 NR_038085.1 Refseq
Affymetrix Mouse Clariom S Array PSR1700024220.mm.1 NR_038086.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR1700024220.mm.1 NR_038086.1 Refseq
Affymetrix Mouse Clariom S Array PSR1700024220.mm.1 TC1700002661.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR1700024221.mm.1 NR_015385.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR1700024221.mm.1 NR_015385.2 Refseq
Affymetrix Mouse Clariom S Array PSR1700024221.mm.1 NR_015386.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR1700024221.mm.1 NR_015386.2 Refseq
Affymetrix Mouse Clariom S Array PSR1700024221.mm.1 NR_015387.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR1700024221.mm.1 NR_015387.2 Refseq
Affymetrix Mouse Clariom S Array PSR1700024221.mm.1 NR_038082.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR1700024221.mm.1 NR_038082.1 Refseq
Affymetrix Mouse Clariom S Array PSR1700024221.mm.1 NR_038083.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR1700024221.mm.1 NR_038083.1 Refseq
Affymetrix Mouse Clariom S Array PSR1700024221.mm.1 NR_038084.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR1700024221.mm.1 NR_038084.1 Refseq
Affymetrix Mouse Clariom S Array PSR1700024221.mm.1 NR_038085.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR1700024221.mm.1 NR_038085.1 Refseq
Affymetrix Mouse Clariom S Array PSR1700024221.mm.1 NR_038086.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR1700024221.mm.1 NR_038086.1 Refseq
Affymetrix Mouse Clariom S Array PSR1700024221.mm.1 TC1700002661.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR1700024215.mm.1 NR_015387.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR1700024215.mm.1 NR_015387.2 Refseq
Affymetrix Mouse Clariom S Array PSR1700024215.mm.1 TC1700002661.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-649bfd9c6b-jlsz7:80/100.66.128.142:80.
git-commit: 6001d82975f0049f9efc09ec70b1b2d89229eeb3
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0