This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm00558032_m1
See other IFNLR1 GE Assays ›
Gene Symbol:
Ifnlr1
Gene Name:
interferon lambda receptor 1
Gene Aliases:
CRF2-12, Il28ra
Chromosome Location:
Chr.4: 135686457 - 135708181 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm00558032_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Ifnlr1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_174851.3 NP_777276.3 1 - 2 143 78
GenBank mRNA AK141338.1 - 1 - 2 144 78
AY184376.1 - 1 - 2 58 78
BC057856.1 - 1 - 2 76 5036481 78

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC0400001661.mm.1

Set Membership:

Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Ifnlr1

Panther Classification:

Molecular Function -

transmembrane signal receptor

Gene Ontology Categories:

Function(s) Process(es)

mucosal immune response
cytokine-mediated signaling pathway
regulation of defense response to virus by host
defense response to virus
cytokine receptor activity

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0400013230.mm.1 NM_174851.3.1 Refseq
Affymetrix Mouse Clariom S Array PSR0400013230.mm.1 NM_174851.3 Refseq
Affymetrix Mouse Clariom S Array PSR0400013230.mm.1 TC0400001661.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0400013229.mm.1 NM_174851.3.1 Refseq
Affymetrix Mouse Clariom S Array PSR0400013229.mm.1 NM_174851.3 Refseq
Affymetrix Mouse Clariom S Array PSR0400013229.mm.1 TC0400001661.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0400013231.mm.1 NM_174851.3.1 Refseq
Affymetrix Mouse Clariom S Array PSR0400013231.mm.1 NM_174851.3 Refseq
Affymetrix Mouse Clariom S Array PSR0400013231.mm.1 TC0400001661.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-56696975f8-skfwn:80/100.66.128.142:80.
git-commit: 795bdb1f794b4b81290d0c522fee4b91276a707f
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0