This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm00557618_m1
See other H6PD GE Assays ›
Gene Symbol:
H6pd
Gene Name:
hexose-6-phosphate dehydrogenase (glucose 1-dehydrogenase)
Gene Aliases:
AI785303, G6pd1, Gpd-1, Gpd1
Chromosome Location:
Chr.4: 149979474 - 150009023 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm00557618_m1
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: H6pd
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
GenBank mRNA AK159373.1 - 4 - 5 1170 66
AK172249.1 - 4 - 5 1176 66
BC042677.1 - 4 - 5 1122 4195793 66

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC0400004058.mm.1

Set Membership:

Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons

Panther Classification:

Molecular Function -

dehydrogenase

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0400033743.mm.1 NM_173371.4 Refseq
Affymetrix Mouse Clariom S Array PSR0400033743.mm.1 TC0400004058.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0400033742.mm.1 NM_173371.4 Refseq
Affymetrix Mouse Clariom S Array PSR0400033742.mm.1 TC0400004058.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0