This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm00554813_m1
See other PCGF3 GE Assays ›
Gene Symbol:
Pcgf3
Gene Name:
polycomb group ring finger 3
Gene Aliases:
2310035N15Rik, AI662857, D630042K08Rik, DONG1, E430039C14, RNF3A, Rnf3
Chromosome Location:
Chr.5: 108461215 - 108503099 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm00554813_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Pcgf3
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_172716.4 NP_766304.2 7 - 8 683 84
GenBank mRNA AK033609.1 - 7 - 8 758 84
AK089091.1 - 7 - 8 684 84
BC049266.1 - 550 5371040 84

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC0500001106.mm.1

Set Membership:

Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Pcgf3

Panther Classification:

Molecular Function -

histone modifying enzyme

Gene Ontology Categories:

Function(s) Process(es)

transcription, DNA-templated
regulation of transcription, DNA-templated
biological_process
molecular_function
zinc ion binding
metal ion binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0500009432.mm.1 NM_172716.4.1 Refseq
Affymetrix Mouse Clariom S Array PSR0500009432.mm.1 NM_172716.4 Refseq
Affymetrix Mouse Clariom S Array PSR0500009432.mm.1 TC0500001106.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0500009433.mm.1 NM_172716.4.1 Refseq
Affymetrix Mouse Clariom S Array PSR0500009433.mm.1 NM_172716.4 Refseq
Affymetrix Mouse Clariom S Array PSR0500009433.mm.1 TC0500001106.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-69fc894774-tvlwx:80/100.66.128.142:80.
git-commit: 8e82defa9bf25b3b77800b5e306013d4ba941391
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0