This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm00553640_g1
See other SERINC5 GE Assays ›
Gene Symbol:
Serinc5
Gene Name:
serine incorporator 5
Gene Aliases:
A130038L21Rik, AIGP3, C86123, TPO1
Chromosome Location:
Chr.13: 92611138 - 92711947 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm00553640_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Serinc5
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_172588.2 NP_766176.1 10 - 11 1223 68
GenBank mRNA AB029501.1 - 10 - 11 1112 68
AK037699.1 - 10 - 11 1223 68
AK054475.1 - 10 - 11 1234 68
AK136799.1 - 10 - 11 1271 68
AK165334.1 - 10 - 11 1230 68
BC049189.1 - 10 - 11 1223 6311617 68
BC062131.1 - 10 - 11 1223 6514513 68

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC1300001036.mm.1

Set Membership:

Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Serinc5

Panther Classification:

Molecular Function -

protein-binding activity modulator

Gene Ontology Categories:

Function(s) Process(es)

lipid metabolic process
phosphatidylserine metabolic process
sphingolipid metabolic process
phospholipid biosynthetic process
L-serine transport
myelination
positive regulation of transferase activity
cation transmembrane transport
L-alpha-amino acid transmembrane transport
molecular_function
L-serine transmembrane transporter activity

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR1300006613.mm.1 NM_172588.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR1300006613.mm.1 NM_172588.2 Refseq
Affymetrix Mouse Clariom S Array PSR1300006613.mm.1 TC1300001036.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR1300006612.mm.1 NM_172588.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR1300006612.mm.1 NM_172588.2 Refseq
Affymetrix Mouse Clariom S Array PSR1300006612.mm.1 TC1300001036.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0