This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm00552942_m1
See other GABPB2 GE Assays ›
Gene Symbol:
Gabpb2
Gene Name:
GA repeat binding protein, beta 2
Gene Aliases:
1810015F01Rik, 5830427M07, 9430006E19Rik, A430024B14Rik, AV050852, Gabpb2-1
Chromosome Location:
Chr.3: 95181766 - 95217951 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm00552942_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Gabpb2
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_029885.1 NP_084161.1 6 - 7 1014 90
NM_172512.2 NP_766100.1 5 - 6 917 90
GenBank mRNA AK030849.1 - 5 - 6 686 90
AK036692.1 - 5 - 6 1498 90
AK141382.1 - 6 - 7 988 90
AK163939.1 - 5 - 6 1494 90
BC150160.1 - 6 - 7 765 40141577 90
BC150482.1 - 6 - 7 765 40141578 90

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC0300002469.mm.1

Set Membership:

Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Gabpb2

Gene Ontology Categories:

Function(s) Process(es)

transcription, DNA-templated
regulation of transcription, DNA-templated
positive regulation of transcription from RNA polymerase II promoter
protein homodimerization activity
transcription regulatory region DNA binding
protein heterodimerization activity

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0300019074.mm.1 NM_029885.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0300019074.mm.1 NM_029885.1 Refseq
Affymetrix Mouse Clariom S Array PSR0300019074.mm.1 NM_172512.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR0300019074.mm.1 NM_172512.2 Refseq
Affymetrix Mouse Clariom S Array PSR0300019074.mm.1 TC0300002469.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0