This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm00552214_m1
See other DDX19B GE Assays ›
Gene Symbol:
Ddx19b
Gene Name:
DEAD (Asp-Glu-Ala-Asp) box polypeptide 19b
Gene Aliases:
2810457M08Rik, 4921519L13Rik, AW260119
Chromosome Location:
Chr.8: 111003186 - 111031751 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm00552214_m1
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Ddx19b
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001190800.1 NP_001177729.1 1 - 2 178 88
NM_172284.5 NP_758488.1 1 - 2 178 88
GenBank mRNA AK147499.1 - 1 - 2 173 88
BC025594.1 - 1 - 2 138 5342859 88

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC0800002935.mm.1

Set Membership:

Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Ddx19b

Gene Ontology Categories:

Function(s) Process(es)

mRNA export from nucleus
biological_process
RNA secondary structure unwinding
molecular_function
ATP-dependent RNA helicase activity

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0800022599.mm.1 NM_001190800.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0800022599.mm.1 NM_001190800.1 Refseq
Affymetrix Mouse Clariom S Array PSR0800022599.mm.1 NM_172284.5.1 Refseq
Affymetrix Mouse Clariom S Array PSR0800022599.mm.1 NM_172284.5 Refseq
Affymetrix Mouse Clariom S Array PSR0800022599.mm.1 TC0800002935.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0800022597.mm.1 NM_001190786.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0800022597.mm.1 NM_001190786.1 Refseq
Affymetrix Mouse Clariom S Array PSR0800022597.mm.1 NM_001190800.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0800022597.mm.1 NM_001190800.1 Refseq
Affymetrix Mouse Clariom S Array PSR0800022597.mm.1 NM_172284.5.1 Refseq
Affymetrix Mouse Clariom S Array PSR0800022597.mm.1 NM_172284.5 Refseq
Affymetrix Mouse Clariom S Array PSR0800022597.mm.1 TC0800002935.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0800022598.mm.1 NM_001190800.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0800022598.mm.1 NM_001190800.1 Refseq
Affymetrix Mouse Clariom S Array PSR0800022598.mm.1 NM_172284.5.1 Refseq
Affymetrix Mouse Clariom S Array PSR0800022598.mm.1 NM_172284.5 Refseq
Affymetrix Mouse Clariom S Array PSR0800022598.mm.1 TC0800002935.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-67c6c7f55f-2g6mz:80/100.66.128.142:80.
git-commit: 92b42c745e7842b39e6d37af9c4064bd81a92e4d
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/search_redesign_header