This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm00552110_m1
See other PDE8B GE Assays ›
Gene Symbol:
Pde8b
Gene Name:
phosphodiesterase 8B
Gene Aliases:
B230331L10Rik, C030047E14Rik
Chromosome Location:
Chr.13: 95024105 - 95250397 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm00552110_m1
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Pde8b
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001170669.1 NP_001164140.1 11 - 12 1476 70
NM_172263.2 NP_758467.2 12 - 13 1209 70
GenBank mRNA AK036090.1 - 14 - 15 1546 70
AK045991.1 - 11 - 12 1478 70
AK083166.1 - 11 - 12 1114 70
BC032286.1 - 12 - 13 1221 5367055 70
EF644173.1 - 14 - 15 1221 70

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC1300002453.mm.1

Set Membership:

Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Pde8b

Panther Classification:

Molecular Function -

phosphodiesterase

Gene Ontology Categories:

Function(s) Process(es)

phosphorelay signal transduction system
behavioral fear response
signal transduction
visual learning
operant conditioning
negative regulation of insulin secretion
neuromuscular process controlling balance
negative regulation of steroid hormone biosynthetic process
3',5'-cyclic-nucleotide phosphodiesterase activity
3',5'-cyclic-AMP phosphodiesterase activity
signal transducer activity
phosphoric diester hydrolase activity
hydrolase activity
metal ion binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR1300016366.mm.1 NM_001170669.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR1300016366.mm.1 NM_001170669.1 Refseq
Affymetrix Mouse Clariom S Array PSR1300016366.mm.1 NM_172263.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR1300016366.mm.1 NM_172263.2 Refseq
Affymetrix Mouse Clariom S Array PSR1300016366.mm.1 TC1300002453.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-4mbwj:80/100.66.130.94:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0