This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm00523323_m1
See other GCKR GE Assays ›
Gene Symbol:
Gckr
Gene Name:
glucokinase regulatory protein
Gene Aliases:
GKRP
Chromosome Location:
Chr.5: 31297254 - 31327302 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm00523323_m1
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Gckr
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_144909.1 NP_659158.1 3 - 4 330 97
GenBank mRNA AK149377.1 - 3 - 4 357 97
BC012412.1 - 3 - 4 330 4159892 97

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC0500000319.mm.1

Set Membership:

Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Gckr

Panther Classification:

Molecular Function -

protein-binding activity modulator

Gene Ontology Categories:

Function(s) Process(es)

protein import into nucleus, translocation
cellular glucose homeostasis
carbohydrate metabolic process
response to fructose
negative regulation of glucokinase activity
positive regulation of glucokinase activity
protein localization to nucleus
negative regulation of catalytic activity
urate metabolic process
triglyceride homeostasis
enzyme inhibitor activity
enzyme binding
protein domain specific binding
carbohydrate binding
fructose-6-phosphate binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0500002822.mm.1 NM_144909.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0500002822.mm.1 NM_144909.1 Refseq
Affymetrix Mouse Clariom S Array PSR0500002822.mm.1 TC0500000319.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-56ffd697dc-88d4p:80/100.66.130.94:80.
git-commit: 082a6d415be982b1f92c35e55dd71609d4576d3e
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.1-2025.09.37-1.0