This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm00523247_m1
See other ATP1A1 GE Assays ›
Gene Symbol:
Atp1a1
Gene Name:
ATPase, Na+/K+ transporting, alpha 1 polypeptide
Gene Aliases:
Atpa-1, BC010319
Chromosome Location:
Chr.3: 101576219 - 101604707 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm00523247_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Atp1a1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_144900.2 NP_659149.1 2 - 3 409 68
GenBank mRNA AK088000.1 - 2 - 3 370 68
AK137001.1 - 2 - 3 368 68
AK159279.1 - 2 - 3 366 68
AK159306.1 - 2 - 3 366 68
AK194265.1 - 1 - 2 78 68
AK195858.1 - 1 - 2 78 68
AK214941.1 - 1 - 2 78 68
BC021496.1 - 2 - 3 359 5346159 68
BC023794.1 - 3 - 4 304 5351943 68
BC023964.1 - 2 - 3 349 68
BC025618.1 - 2 - 3 368 5320976 68
BC025627.1 - 2 - 3 359 5320513 68
BC025811.1 - 2 - 3 346 5096872 68
BC032187.1 - 2 - 3 409 5321268 68
BC033435.1 - 2 - 3 305 4950746 68
BC033471.1 - 2 - 3 349 4954119 68
BC034177.1 - 1 - 2 114 68
BC035352.1 - 1 - 2 79 68
BC042435.1 - 2 - 3 359 4951288 68

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC0300002605.mm.1

Set Membership:

Ambion siRNA available Amplicon length less than or equal to 70 Ion Channel Transporter Mammalian Gene Collection Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Atp1a1

Panther Classification:

Molecular Function -

primary active transporter

Gene Ontology Categories:

Function(s) Process(es)

regulation of the force of heart contraction
regulation of sodium ion transport
transport
ion transport
potassium ion transport
sodium ion transport
cellular sodium ion homeostasis
regulation of blood pressure
potassium ion import
dephosphorylation
cellular potassium ion homeostasis
negative regulation of glucocorticoid biosynthetic process
sodium ion transmembrane transport
sodium ion export from cell
response to drug
negative regulation of heart contraction
positive regulation of heart contraction
positive regulation of striated muscle contraction
membrane hyperpolarization
cellular response to mechanical stimulus
cellular response to steroid hormone stimulus
potassium ion transmembrane transport
regulation of cardiac muscle cell contraction
membrane repolarization
response to glycoside
potassium ion import across plasma membrane
nucleotide binding
sodium:potassium-exchanging ATPase activity
protein binding
ATP binding
hydrolase activity
phosphatase activity
protein kinase binding
protein domain specific binding
ankyrin binding
potassium ion binding
sodium ion binding
ADP binding
phosphatidylinositol 3-kinase binding
metal ion binding
chaperone binding
steroid hormone binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0