This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm00521889_m1
See other LARGE GE Assays ›
Gene Symbol:
Large
Gene Name:
like-glycosyltransferase
Gene Aliases:
BPFD#36, Gyltl1a, Mbp-1, Mbp1, enr, fg, froggy, mKIAA0609, myd
Chromosome Location:
Chr.8: 72814599 - 73353577 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm00521889_m1
Size
Availability Made To Order
Catalog # 4351372
Price 4,408.00
Your Price
Online offer:
4408.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Large
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_010687.1 NP_034817.1 8 - 9 1176 92
GenBank mRNA AJ006278.1 - 8 - 9 1176 92
AK122328.1 - 9 - 10 2253 92
AK133940.1 - 8 - 9 1660 92
BC061506.1 - 7 - 8 933 92
BC079596.1 - 8 - 9 1459 92
BC125487.1 - 8 - 9 1142 40130002 92
BC125489.1 - 8 - 9 1142 40130004 92
HM141820.1 - 6 - 7 811 92

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC0800002471.mm.1

Set Membership:

Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Large

Panther Classification:

Molecular Function -

glycosyltransferase

Gene Ontology Categories:

Function(s) Process(es)

metabolic process
glycoprotein biosynthetic process
protein O-linked mannosylation
skeletal muscle tissue regeneration
muscle cell cellular homeostasis
skeletal muscle organ development
alpha-1,6-mannosyltransferase activity
alpha-1,2-mannosyltransferase activity
mannosyltransferase activity
alpha-1,3-mannosyltransferase activity
imidazoleglycerol-phosphate synthase activity
alpha-1,3-galactosyltransferase activity
catalytic activity
UDP-glucose:glycoprotein glucosyltransferase activity
glycolipid mannosyltransferase activity
oligosaccharyl transferase activity
dolichyl-phosphate-glucose-glycolipid alpha-glucosyltransferase activity
protein binding
acetylglucosaminyltransferase activity
acetylgalactosaminyltransferase activity
galactosyltransferase activity
fucosyltransferase activity
O antigen polymerase activity
lipopolysaccharide-1,6-galactosyltransferase activity
glucuronosyltransferase activity
transferase activity
transferase activity, transferring glycosyl groups
cellulose synthase activity
NAD(P)-cysteine ADP-ribosyltransferase activity
NAD(P)-asparagine ADP-ribosyltransferase activity
NAD(P)-serine ADP-ribosyltransferase activity
9-phenanthrol UDP-glucuronosyltransferase activity
1-phenanthrol glycosyltransferase activity
9-phenanthrol glycosyltransferase activity
1,2-dihydroxy-phenanthrene glycosyltransferase activity
phenanthrol glycosyltransferase activity
beta-1,4-mannosyltransferase activity
manganese ion binding
alpha-1,2-galactosyltransferase activity
dolichyl pyrophosphate Man7GlcNAc2 alpha-1,3-glucosyltransferase activity
lipopolysaccharide-1,5-galactosyltransferase activity
dolichyl pyrophosphate Man9GlcNAc2 alpha-1,3-glucosyltransferase activity
dolichyl pyrophosphate Glc1Man9GlcNAc2 alpha-1,3-glucosyltransferase activity
xylosyltransferase activity
7-cyano-7-deazaguanine tRNA-ribosyltransferase activity
purine deoxyribosyltransferase activity
inositol phosphoceramide synthase activity
glucosyltransferase activity
metal ion binding
alpha-(1->3)-fucosyltransferase activity
alpha-(1->6)-fucosyltransferase activity
indole-3-butyrate beta-glucosyltransferase activity
salicylic acid glucosyltransferase (ester-forming) activity
salicylic acid glucosyltransferase (glucoside-forming) activity
benzoic acid glucosyltransferase activity
chondroitin hydrolase activity
dolichyl-pyrophosphate Man7GlcNAc2 alpha-1,6-mannosyltransferase activity
cytokinin 9-beta-glucosyltransferase activity

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0800018919.mm.1 NM_010687.2 Refseq
Affymetrix Mouse Clariom S Array PSR0800018919.mm.1 TC0800002471.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-649bfd9c6b-jlsz7:80/100.66.128.142:80.
git-commit: 6001d82975f0049f9efc09ec70b1b2d89229eeb3
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0