This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm00521608_m1
See other FAM129B GE Assays ›
Gene Symbol:
Fam129b
Gene Name:
family with sequence similarity 129, member B
Gene Aliases:
9130404D14Rik
Chromosome Location:
Chr.2: 32876134 - 32925253 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm00521608_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Fam129b
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_146119.2 NP_666231.1 9 - 10 1372 67
GenBank mRNA AK033735.1 - 9 - 10 1340 67
AK046583.1 - 9 - 10 1269 67
AK051396.1 - 9 - 10 1311 67
AK146953.1 - 9 - 10 1357 67
AK149818.1 - 9 - 10 1283 67
AK149888.1 - 9 - 10 1282 67
AK150642.1 - 9 - 10 1310 67
AK152207.1 - 9 - 10 1331 67
AK153489.1 - 9 - 10 1316 67
AK170512.1 - 9 - 10 1312 67
AK178139.1 - 1 - 2 32 67
BC024639.1 - 1 - 2 105 67
BC027843.1 - 9 - 10 1292 5343013 67

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC0200000597.mm.1

Set Membership:

Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Fam129b

Gene Ontology Categories:

Function(s)

negative regulation of apoptotic process

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0200005508.mm.1 NM_146119.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR0200005508.mm.1 NM_146119.2 Refseq
Affymetrix Mouse Clariom S Array PSR0200005508.mm.1 TC0200000597.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0200005509.mm.1 NM_146119.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR0200005509.mm.1 NM_146119.2 Refseq
Affymetrix Mouse Clariom S Array PSR0200005509.mm.1 TC0200000597.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-69fc894774-tvlwx:80/100.66.128.142:80.
git-commit: 8e82defa9bf25b3b77800b5e306013d4ba941391
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0