This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm00521602_m1
See other FAM129B GE Assays ›
Gene Symbol:
Fam129b
Gene Name:
family with sequence similarity 129, member B
Gene Aliases:
9130404D14Rik
Chromosome Location:
Chr.2: 32876134 - 32925253 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm00521602_m1
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Fam129b
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_146119.2 NP_666231.1 3 - 4 536 78
GenBank mRNA AK033735.1 - 3 - 4 504 78
AK046583.1 - 3 - 4 433 78
AK051396.1 - 3 - 4 475 78
AK146953.1 - 3 - 4 521 78
AK149818.1 - 3 - 4 447 78
AK149888.1 - 3 - 4 446 78
AK150642.1 - 3 - 4 474 78
AK152207.1 - 3 - 4 495 78
AK153489.1 - 3 - 4 480 78
AK170512.1 - 3 - 4 476 78
BC027843.1 - 3 - 4 456 5343013 78

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC0200000597.mm.1

Set Membership:

Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Fam129b

Gene Ontology Categories:

Function(s)

negative regulation of apoptotic process

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0200005497.mm.1 NM_146119.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR0200005497.mm.1 NM_146119.2 Refseq
Affymetrix Mouse Clariom S Array PSR0200005497.mm.1 TC0200000597.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0