This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm00521014_m1
See other ME2 GE Assays ›
Gene Symbol:
Me2
Gene Name:
malic enzyme 2, NAD(+)-dependent, mitochondrial
Gene Aliases:
AW120568, D030040L20Rik, NAD-ME
Chromosome Location:
Chr.18: 73764956 - 73815478 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm00521014_m1
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Me2
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_145494.2 NP_663469.1 1 - 2 89 75
GenBank mRNA AK039906.1 - 1 - 2 136 75
AK042289.1 - 1 - 2 136 75
AK043103.1 - 1 - 2 127 75
AK050933.1 - 1 - 2 136 75
AK050980.1 - 1 - 2 136 75
AK077746.1 - 1 - 2 136 75
AK080403.1 - 1 - 2 147 75
AK156403.1 - 1 - 2 136 75
AK162126.1 - 1 - 2 148 75
AK171157.1 - 1 - 2 136 75
BC004709.1 - 1 - 2 89 3499365 75

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC1800001581.mm.1

Additional Information:

This assay is in the 5'UTR region of the AK042289.1,AK050933.1,AK050980.1,AK080403.1,AK156403.1,AK162126.1,AK171157.1,BC004709.1,NM_145494.2 transcripts.

Set Membership:

Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus 1700 Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Me2

Panther Classification:

Molecular Function -

oxidoreductase

Gene Ontology Categories:

Function(s) Process(es)

malate metabolic process
metabolic process
oxidation-reduction process
regulation of NADP metabolic process
catalytic activity
malic enzyme activity
malate dehydrogenase (decarboxylating) (NAD+) activity
oxaloacetate decarboxylase activity
oxidoreductase activity
metal ion binding
NAD binding

MicroArray Probes:

Affymetrix ABI

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR1800011448.mm.1 NM_145494.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR1800011448.mm.1 NM_145494.2 Refseq
Affymetrix Mouse Clariom S Array PSR1800011448.mm.1 TC1800001581.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR1800011446.mm.1 NM_145494.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR1800011446.mm.1 NM_145494.2 Refseq
Affymetrix Mouse Clariom S Array PSR1800011446.mm.1 TC1800001581.mm.1 MTA TC
Array Name Probe IDs Targets Target Type
ABI Mouse Genome Survey Microarray 522483 - -

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0