This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm00520903_m1
See other PXK GE Assays ›
Gene Symbol:
Pxk
Gene Name:
PX domain containing serine/threonine kinase
Gene Aliases:
C230080L11Rik, D14Ertd813e, MONaKA
Chromosome Location:
Chr.14: 8098191 - 8165117 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm00520903_m1
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Pxk
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_145458.3 NP_663433.2 13 - 14 1327 101
NM_178279.2 NP_840063.1 13 - 14 1327 101
GenBank mRNA AK028501.1 - 13 - 14 1277 101
AK048910.1 - 13 - 14 1287 101
AK147037.1 - 13 - 14 1327 101
AK150337.1 - 13 - 14 1312 101
AK150413.1 - 13 - 14 1312 101
AK150660.1 - 13 - 14 1328 101
AK151098.1 - 13 - 14 1288 101
AK152058.1 - 13 - 14 1286 101
AK152789.1 - 13 - 14 1286 101
AK153067.1 - 13 - 14 1286 101
AK156146.1 - 13 - 14 1293 101
AK157364.1 - 13 - 14 1321 101
AK166036.1 - 13 - 14 1253 101
AK169112.1 - 13 - 14 1287 101
AK170361.1 - 13 - 14 1323 101
AK172463.1 - 13 - 14 1329 101
BC016131.1 - 13 - 14 1297 4922274 101
DQ095196.1 - 13 - 14 1225 101
DQ095197.1 - 13 - 14 1225 101

Back To Top

More Information


Set Membership:

Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Pxk

Gene Ontology Categories:

Function(s) Process(es)

protein phosphorylation
inflammatory response
negative regulation of ATPase activity
negative regulation of ion transport
actin binding
protein kinase activity
ATP binding
protein C-terminus binding
transferase activity, transferring phosphorus-containing groups
phosphatidylinositol binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0