This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm00520708_m1
See other NR1D1 GE Assays ›
Gene Symbol:
Nr1d1
Gene Name:
nuclear receptor subfamily 1, group D, member 1
Gene Aliases:
A530070C09Rik, R75201
Chromosome Location:
Chr.11: 98767932 - 98775377 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm00520708_m1
Best Coverage
Size
Availability Inventoried
Catalog # 4331182
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Nr1d1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_145434.4 NP_663409.2 1 - 2 664 62
GenBank mRNA AF291821.1 - 1 - 2 488 62
AK041047.1 - 1 - 2 519 62
AK135640.1 - 1 - 2 526 62
AK137398.1 - 1 - 2 551 62
AK137582.1 - 1 - 2 529 62
AK146430.1 - 1 - 2 621 62
AK154931.1 - 1 - 2 665 62
AK155597.1 - 1 - 2 665 62
BC008989.1 - 1 - 2 109 3585478 62
X86010.1 - 1 - 2 524 62

Back To Top

More Information


Set Membership:

Ambion siRNA available Amplicon length less than or equal to 70 Nuclear Receptor Mammalian Gene Collection Recommended Assay Mus musculus 1700 Inventoried Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Nr1d1

Panther Classification:

Molecular Function -

C4 zinc finger nuclear receptor DNA-binding transcription factor zinc finger transcription factor gene-specific transcriptional regulator

Gene Ontology Categories:

Function(s) Process(es)

negative regulation of transcription from RNA polymerase II promoter
glycogen biosynthetic process
transcription, DNA-templated
regulation of transcription, DNA-templated
transcription from RNA polymerase II promoter
circadian rhythm
proteasomal protein catabolic process
negative regulation of receptor biosynthetic process
regulation of lipid metabolic process
cell differentiation
circadian regulation of gene expression
negative regulation of toll-like receptor 4 signaling pathway
regulation of gluconeogenesis by regulation of transcription from RNA polymerase II promoter
regulation of circadian rhythm
steroid hormone mediated signaling pathway
response to leptin
regulation of fat cell differentiation
negative regulation of transcription, DNA-templated
positive regulation of transcription, DNA-templated
rhythmic process
circadian temperature homeostasis
regulation of insulin secretion involved in cellular response to glucose stimulus
regulation of type B pancreatic cell proliferation
positive regulation of bile acid biosynthetic process
cellular response to lipopolysaccharide
regulation of cholesterol homeostasis
RNA polymerase II regulatory region sequence-specific DNA binding
RNA polymerase II core promoter proximal region sequence-specific DNA binding
core promoter sequence-specific DNA binding
transcriptional repressor activity, RNA polymerase II core promoter proximal region sequence-specific binding
transcription corepressor binding
DNA binding
transcription factor activity, sequence-specific DNA binding
steroid hormone receptor activity
protein binding
zinc ion binding
heme binding
sequence-specific DNA binding
transcription regulatory region DNA binding
metal ion binding

MicroArray Probes:

ABI

Array Name Probe IDs Targets Target Type
ABI Mouse Genome Survey Microarray 650526 - -

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0