This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm00520665_m1
See other ARRB2 GE Assays ›
Gene Symbol:
Arrb2
Gene Name:
arrestin, beta 2
Gene Aliases:
AI326910, AW122872
Chromosome Location:
Chr.11: 70432580 - 70440828 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm00520665_m1
Size
Availability Inventoried
Catalog # 4331182
Price 244.00
Your Price
Online offer: 200.00
200.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Arrb2
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001271358.1 NP_001258287.1 1 - 2 180 70
NM_001271359.1 NP_001258288.1 1 - 2 180 70
NM_001271360.1 NP_001258289.1 1 - 2 180 70
NM_145429.5 NP_663404.1 1 - 2 180 70
GenBank mRNA AK089542.1 - 1 - 2 115 70
AK150389.1 - 1 - 2 157 70
AK154874.1 - 1 - 2 100 70
AK159317.1 - 1 - 2 108 70
AK162889.1 - 1 - 2 117 70
AK170889.1 - 1 - 2 90 70
BC016642.1 - 1 - 2 2651372 70
BY201367.1 - 1 - 2 180 70

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC1100000950.mm.1

Additional Information:

This assay is in the 5'UTR region of the AK150389.1,AK154874.1,AK159317.1,AK162889.1,AK170889.1,BC016642.1,NM_001271358.1,NM_001271359.1,NM_001271360.1,NM_145429.5 transcripts.

Set Membership:

Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus 1700 Inventoried Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Arrb2

Panther Classification:

Molecular Function -

scaffold/adaptor protein

Gene Ontology Categories:

Function(s) Process(es)

regulation of protein phosphorylation
positive regulation of protein phosphorylation
G-protein coupled receptor internalization
desensitization of G-protein coupled receptor protein signaling pathway by arrestin
positive regulation of receptor internalization
transcription from RNA polymerase II promoter
transport
endocytosis
signal transduction
transforming growth factor beta receptor signaling pathway
G-protein coupled receptor signaling pathway
adult walking behavior
regulation of G-protein coupled receptor protein signaling pathway
negative regulation of signal transduction
protein transport
protein ubiquitination
negative regulation of protein ubiquitination
positive regulation of protein ubiquitination
receptor internalization
negative regulation of NF-kappaB transcription factor activity
positive regulation of synaptic transmission, dopaminergic
negative regulation of interleukin-1 beta production
negative regulation of interleukin-12 production
negative regulation of interleukin-6 production
negative regulation of tumor necrosis factor production
positive regulation of peptidyl-serine phosphorylation
negative regulation of toll-like receptor signaling pathway
negative regulation of GTPase activity
negative regulation of smooth muscle cell apoptotic process
follicle-stimulating hormone signaling pathway
negative regulation of cysteine-type endopeptidase activity involved in apoptotic process
proteasome-mediated ubiquitin-dependent protein catabolic process
negative regulation of natural killer cell mediated cytotoxicity
positive regulation of peptidyl-tyrosine phosphorylation
detection of temperature stimulus involved in sensory perception of pain
positive regulation of protein kinase B signaling
positive regulation of calcium ion transport
cell chemotaxis
regulation of androgen receptor signaling pathway
positive regulation of ERK1 and ERK2 cascade
negative regulation of release of cytochrome c from mitochondria
positive regulation of DNA biosynthetic process
G-protein coupled receptor binding
receptor binding
protein binding
enzyme binding
protein domain specific binding
ubiquitin protein ligase binding
alpha-1A adrenergic receptor binding
alpha-1B adrenergic receptor binding
angiotensin receptor binding
type 1 angiotensin receptor binding
D1 dopamine receptor binding
follicle-stimulating hormone receptor binding
type 2A serotonin receptor binding
platelet activating factor receptor binding
protein complex binding
protein complex scaffold
protein kinase B binding
mitogen-activated protein kinase binding
14-3-3 protein binding

MicroArray Probes:

Affymetrix ABI

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR1100008311.mm.1 NM_001271358.1 Refseq
Affymetrix Mouse Clariom S Array PSR1100008311.mm.1 NM_001271359.1 Refseq
Affymetrix Mouse Clariom S Array PSR1100008311.mm.1 NM_001271360.1 Refseq
Affymetrix Mouse Clariom S Array PSR1100008311.mm.1 NM_145429.5 Refseq
Affymetrix Mouse Clariom S Array PSR1100008311.mm.1 TC1100000950.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR1100008321.mm.1 NM_001271358.1 Refseq
Affymetrix Mouse Clariom S Array PSR1100008321.mm.1 NM_001271359.1 Refseq
Affymetrix Mouse Clariom S Array PSR1100008321.mm.1 NM_001271360.1 Refseq
Affymetrix Mouse Clariom S Array PSR1100008321.mm.1 NM_145429.5 Refseq
Affymetrix Mouse Clariom S Array PSR1100008321.mm.1 TC1100000950.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR1100008318.mm.1 NM_001271358.1 Refseq
Affymetrix Mouse Clariom S Array PSR1100008318.mm.1 NM_001271359.1 Refseq
Affymetrix Mouse Clariom S Array PSR1100008318.mm.1 NM_001271360.1 Refseq
Affymetrix Mouse Clariom S Array PSR1100008318.mm.1 NM_145429.5 Refseq
Affymetrix Mouse Clariom S Array PSR1100008318.mm.1 TC1100000950.mm.1 MTA TC
Array Name Probe IDs Targets Target Type
ABI Mouse Genome Survey Microarray 854878 - -

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0