This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm00520173_m1
See other POMT1 GE Assays ›
Gene Symbol:
Pomt1
Gene Name:
protein-O-mannosyltransferase 1
Gene Aliases:
AI505244
Chromosome Location:
Chr.2: 32236595 - 32255005 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm00520173_m1
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Pomt1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_145145.1 NP_660127.1 5 - 6 520 54
GenBank mRNA AK053889.1 - 5 - 6 519 54
AK134770.1 - 5 - 6 564 54
AK153984.1 - 5 - 6 619 54
AY007238.1 - 5 - 6 560 54
AY494857.1 - 5 - 6 580 54
BC027325.1 - 5 - 6 520 3981366 54

Back To Top

More Information


Additional Information:

This assay is in the 5'UTR region of the AK053889.1 transcripts.

Set Membership:

Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Pomt1

Panther Classification:

Molecular Function -

transferase metabolite interconversion enzyme

Gene Ontology Categories:

Function(s) Process(es)

protein O-linked glycosylation
extracellular matrix organization
mannosyltransferase activity
dolichyl-phosphate-mannose-protein mannosyltransferase activity
transferase activity
transferase activity, transferring glycosyl groups

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0