This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm00517261_m1
See other DNAJC7 GE Assays ›
Gene Symbol:
Dnajc7
Gene Name:
DnaJ (Hsp40) homolog, subfamily C, member 7
Gene Aliases:
2010003F24Rik, 2010004G07Rik, CCRP, Ttc2, mDj11, mTpr2
Chromosome Location:
Chr.11: 100582818 - 100620168 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm00517261_m1
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Dnajc7
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_019795.4 NP_062769.2 4 - 5 964 110
GenBank mRNA AB028861.1 - 4 - 5 412 110
AK011875.1 - 4 - 5 403 110
AK076032.1 - 4 - 5 435 110
AK145506.1 - 4 - 5 426 110
AK145739.1 - 4 - 5 426 110
AK166872.1 - 4 - 5 426 110
BC023681.1 - 4 - 5 608 5343384 110
BC055729.1 - 4 - 5 963 6414499 110

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC1100003776.mm.1

Set Membership:

Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Dnajc7

Panther Classification:

Molecular Function -

chaperone

Gene Ontology Categories:

Function(s) Process(es)

chaperone cofactor-dependent protein refolding
heat shock protein binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR1100034755.mm.1 NM_019795.4.1 Refseq
Affymetrix Mouse Clariom S Array PSR1100034755.mm.1 NM_019795.4 Refseq
Affymetrix Mouse Clariom S Array PSR1100034755.mm.1 TC1100003776.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0