This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm00517043_m1
See other GRK5 GE Assays ›
Gene Symbol:
Grk5
Gene Name:
G protein-coupled receptor kinase 5
Gene Aliases:
Gprk5
Chromosome Location:
Chr.19: 60889593 - 61095968 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm00517043_m1
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Grk5
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_018869.3 NP_061357.3 12 - 13 2101 61
GenBank mRNA AF040746.1 - 12 - 13 1511 61
AK135076.1 - 12 - 13 2103 61
AK156465.1 - 11 - 12 2476 61
AK161821.1 - 12 - 13 1581 61
BC019379.1 - 12 - 13 1374 5116510 61

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC1900000866.mm.1

Set Membership:

Ambion siRNA available Amplicon length less than or equal to 70 Protein Kinase Mammalian Gene Collection Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Grk5

Panther Classification:

Molecular Function -

non-receptor serine/threonine protein kinase

Gene Ontology Categories:

Function(s) Process(es)

desensitization of G-protein coupled receptor protein signaling pathway
protein phosphorylation
apoptotic process
signal transduction
tachykinin receptor signaling pathway
regulation of G-protein coupled receptor protein signaling pathway
positive regulation of cell proliferation
Wnt signaling pathway
phosphorylation
termination of G-protein coupled receptor signaling pathway
negative regulation of apoptotic process
protein autophosphorylation
regulation of cell cycle
nucleotide binding
protein kinase activity
protein serine/threonine kinase activity
G-protein coupled receptor kinase activity
ATP binding
lipid binding
kinase activity
transferase activity
transferase activity, transferring phosphorus-containing groups
beta-adrenergic receptor kinase activity

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR1900007484.mm.1 NM_018869.3.1 Refseq
Affymetrix Mouse Clariom S Array PSR1900007484.mm.1 NM_018869.3 Refseq
Affymetrix Mouse Clariom S Array PSR1900007484.mm.1 TC1900000866.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0