This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm00515615_m1
See other CLIP2 GE Assays ›
Gene Symbol:
Clip2
Gene Name:
CAP-GLY domain containing linker protein 2
Gene Aliases:
B230327O20, CLIP-115, Clip1, Cyln2, WSCR4, mKIAA0291, wbscr4
Chromosome Location:
Chr.5: 134489383 - 134552468 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm00515615_m1
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Clip2
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001039162.2 NP_001034251.1 9 - 10 2777 107
NM_009990.3 NP_034120.2 10 - 11 2882 107
GenBank mRNA AK045967.1 - 10 - 11 2727 107
AK172928.1 - 10 - 11 2627 107
BC039162.1 - 9 - 10 2717 3498067 107
BC053048.1 - 9 - 10 2777 5709279 107
BC121191.1 - 9 - 10 2423 107

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC0500003373.mm.1

Set Membership:

Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Clip2

Panther Classification:

Molecular Function -

chaperone

Gene Ontology Categories:

Process(es)

protein binding
microtubule plus-end binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0500030412.mm.1 NM_001039162.2 Refseq
Affymetrix Mouse Clariom S Array PSR0500030412.mm.1 NM_009990.3 Refseq
Affymetrix Mouse Clariom S Array PSR0500030412.mm.1 TC0500003373.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0