This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm00514389_m1
See other CRMP1 GE Assays ›
Gene Symbol:
Crmp1
Gene Name:
collapsin response mediator protein 1
Gene Aliases:
CRMP-1, DRP-1, Dpysl1, ULIP-3, Ulip3
Chromosome Location:
Chr.5: 37242058 - 37292163 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm00514389_m1
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Crmp1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001136058.2 NP_001129530.1 11 - 12 1712 67
NM_007765.4 NP_031791.3 11 - 12 1577 67
GenBank mRNA AB006714.1 - 11 - 12 1549 67
AK159049.1 - 11 - 12 1521 67
BC031738.1 - 11 - 12 1506 4505284 67
BC065046.1 - 11 - 12 1690 5686818 67
U72875.1 - 11 - 12 1322 67
Y09080.1 - 11 - 12 1552 67

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC0500000422.mm.1

Set Membership:

Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Crmp1

Panther Classification:

Molecular Function -

hydrolase

Gene Ontology Categories:

Process(es)

hydrolase activity
hydrolase activity, acting on carbon-nitrogen (but not peptide) bonds
phosphoprotein binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0500004201.mm.1 NM_001136058.2 Refseq
Affymetrix Mouse Clariom S Array PSR0500004201.mm.1 NM_007765.4 Refseq
Affymetrix Mouse Clariom S Array PSR0500004201.mm.1 TC0500000422.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0500004200.mm.1 NM_001136058.2 Refseq
Affymetrix Mouse Clariom S Array PSR0500004200.mm.1 NM_007765.4 Refseq
Affymetrix Mouse Clariom S Array PSR0500004200.mm.1 TC0500000422.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0