This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm00512560_m1
See other SF3A3 GE Assays ›
Gene Symbol:
Sf3a3
Gene Name:
splicing factor 3a, subunit 3
Gene Aliases:
4930512K19Rik, 60kDa
Chromosome Location:
Chr.4: 124714861 - 124732423 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm00512560_m1
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Sf3a3
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_029157.3 NP_083433.1 14 - 15 1318 98
GenBank mRNA AK030743.1 - 14 - 15 1353 98
AK150733.1 - 14 - 15 1316 98
AK151141.1 - 14 - 15 1316 98
AK151382.1 - 14 - 15 1316 98
AK161646.1 - 14 - 15 1339 98
AK168966.1 - 14 - 15 1342 98
AK169324.1 - 14 - 15 1362 98
BC009141.1 - 14 - 15 1318 3598647 98
BC092058.1 - 14 - 15 1350 4017388 98

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC0400001421.mm.1

Set Membership:

Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Sf3a3

Panther Classification:

Molecular Function -

RNA splicing factor

Gene Ontology Categories:

Function(s) Process(es)

mRNA splicing, via spliceosome
mRNA processing
RNA splicing
nucleic acid binding
RNA binding
zinc ion binding
poly(A) RNA binding
metal ion binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0400010690.mm.1 NM_029157.3.1 Refseq
Affymetrix Mouse Clariom S Array PSR0400010690.mm.1 NM_029157.3 Refseq
Affymetrix Mouse Clariom S Array PSR0400010690.mm.1 TC0400001421.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0