This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm00512288_m1
See other TC2N GE Assays ›
Gene Symbol:
Tc2n
Gene Name:
tandem C2 domains, nuclear
Gene Aliases:
4933406D09Rik, Mtac2d1, Tac2-N
Chromosome Location:
Chr.12: 101645446 - 101719000 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm00512288_m1
Size
Availability Made To Order
Catalog # 4351372
Price 4,408.00
Your Price
Online offer:
4408.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Tc2n
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001082976.2 NP_001076445.1 6 - 7 808 111
NM_001286364.1 NP_001273293.1 6 - 7 1471 111
NM_028924.3 NP_083200.1 6 - 7 1160 111
GenBank mRNA AB062282.1 - 5 - 6 640 111
AK016683.1 - 6 - 7 1492 111
BC120793.1 - 6 - 7 1160 40129716 111

Back To Top

More Information


Set Membership:

Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Tc2n

Gene Ontology Categories:

Function(s) Process(es)

regulation of gene expression
regulation of blood coagulation
molecular_function
calcium ion binding
calcium-dependent phospholipid binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-649bfd9c6b-jlsz7:80/100.66.128.142:80.
git-commit: 6001d82975f0049f9efc09ec70b1b2d89229eeb3
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0