This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm00512097_m1
See other SLC35F5 GE Assays ›
Gene Symbol:
Slc35f5
Gene Name:
solute carrier family 35, member F5
Gene Aliases:
1300003P13Rik, AI646727
Chromosome Location:
Chr.1: 125560414 - 125595780 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm00512097_m1
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Slc35f5
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_028787.4 NP_083063.2 9 - 10 1150 92
GenBank mRNA AK004892.2 - 9 - 10 1131 92
AK076339.1 - 9 - 10 1144 92
AK145848.1 - 9 - 10 1151 92
AK169012.1 - 9 - 10 1012 92
AK171951.1 - 9 - 10 1124 92
BC026858.1 - 9 - 10 1009 4014064 92

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC0100001111.mm.1

Set Membership:

Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Slc35f5

Panther Classification:

Molecular Function -

secondary carrier transporter

Gene Ontology Categories:

Function(s) Process(es)

transport
biological_process
molecular_function

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0100009025.mm.1 NM_028787.4.1 Refseq
Affymetrix Mouse Clariom S Array PSR0100009025.mm.1 NM_028787.4 Refseq
Affymetrix Mouse Clariom S Array PSR0100009025.mm.1 TC0100001111.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0