This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm00509311_m1
See other GPR89 GE Assays ›
Gene Symbol:
Gpr89
Gene Name:
G protein-coupled receptor 89
Gene Aliases:
4933412D19Rik, Gpr89a, SH120
Chromosome Location:
Chr.3: 96871066 - 96905343 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm00509311_m1
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Gpr89
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_026229.4 NP_080505.1 6 - 7 606 68
GenBank mRNA AK002937.1 - 6 - 7 626 68
AK009734.1 - 5 - 6 485 68
AK016787.1 - 6 - 7 606 68
AK034931.1 - 174 68
BC010729.1 - 6 - 7 599 3708143 68

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC0300002531.mm.1

Set Membership:

Ambion siRNA available Amplicon length less than or equal to 70 Protein Kinase Mammalian Gene Collection Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Gpr89

Panther Classification:

Molecular Function -

G-protein coupled receptor transmembrane signal receptor

Gene Ontology Categories:

Function(s) Process(es)

transport
ion transport
signal transduction
protein transport
regulation of ion transmembrane transport
skin development
anion transmembrane transport
regulation of anion transmembrane transport
signal transducer activity
voltage-gated ion channel activity
voltage-gated anion channel activity

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0300019738.mm.1 NM_026229.4.1 Refseq
Affymetrix Mouse Clariom S Array PSR0300019738.mm.1 NM_026229.4 Refseq
Affymetrix Mouse Clariom S Array PSR0300019738.mm.1 TC0300002531.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-67c6c7f55f-2g6mz:80/100.66.128.142:80.
git-commit: 92b42c745e7842b39e6d37af9c4064bd81a92e4d
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/search_redesign_header