This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm00508507_m1
See other TIMM50 GE Assays ›
Gene Symbol:
Timm50
Gene Name:
translocase of inner mitochondrial membrane 50
Gene Aliases:
2810403L02Rik, AU015082, TIM50L
Chromosome Location:
Chr.7: 28305516 - 28312046 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm00508507_m1
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Timm50
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_025616.3 NP_079892.1 1 - 2 121 124
GenBank mRNA AK008340.1 - 1 - 2 122 124
AK008382.1 - 1 - 2 119 122
AK012977.1 - 1 - 2 118 123
AK088074.1 - 1 - 2 114 122
AK167473.1 - 1 - 2 124 124
AY551342.1 - 1 - 2 107 124
BC037056.1 - 1 - 2 124 124

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC0700002693.mm.1

Set Membership:

Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus 1700 Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Timm50

Gene Ontology Categories:

Function(s) Process(es)

release of cytochrome c from mitochondria
protein dephosphorylation
transport
mitochondrial membrane organization
protein transport
dephosphorylation
peptidyl-tyrosine dephosphorylation
phosphoprotein phosphatase activity
protein serine/threonine phosphatase activity
protein tyrosine phosphatase activity
interleukin-2 receptor binding
ribonucleoprotein complex binding

MicroArray Probes:

Affymetrix ABI

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0700023523.mm.1 NM_025616.3.1 Refseq
Affymetrix Mouse Clariom S Array PSR0700023523.mm.1 NM_025616.3 Refseq
Affymetrix Mouse Clariom S Array PSR0700023523.mm.1 TC0700002693.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0700023520.mm.1 NM_025616.3.1 Refseq
Affymetrix Mouse Clariom S Array PSR0700023520.mm.1 NM_025616.3 Refseq
Affymetrix Mouse Clariom S Array PSR0700023520.mm.1 TC0700002693.mm.1 MTA TC
Array Name Probe IDs Targets Target Type
ABI Mouse Genome Survey Microarray 918832 - -

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0