This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm00507926_m1
See other ANXA8 GE Assays ›
Gene Symbol:
Anxa8
Gene Name:
annexin A8
Gene Aliases:
AI466995, Anx8
Chromosome Location:
Chr.14: 34085979 - 34100571 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm00507926_m1
Size
Availability Inventoried
Catalog # 4331182
Price 244.00
Your Price
Online offer: 200.00
200.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Anxa8
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001281845.1 NP_001268774.1 1 - 2 95 93
NM_013473.4 NP_038501.2 1 - 2 95 93
GenBank mRNA AJ002390.1 - 1 - 2 71 93
AK009085.1 - 1 - 2 57 93
BC030407.1 - 1 - 2 108 5322310 93

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC1400000368.mm.1

Additional Information:

This assay is in the 5'UTR region of the AJ002390.1,BC030407.1,NM_001281845.1,NM_013473.4 transcripts.

Set Membership:

Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Inventoried Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Anxa8

Panther Classification:

Molecular Function -

calcium-binding protein

Gene Ontology Categories:

Function(s) Process(es)

blood coagulation
hemostasis
calcium ion binding
calcium-dependent phospholipid binding
phosphatidylinositol-4,5-bisphosphate binding
phosphatidylinositol-3,4,5-trisphosphate binding
phosphatidylinositol-3,4-bisphosphate binding
metal ion binding
actin filament binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR1400003283.mm.1 NM_001281845.1 Refseq
Affymetrix Mouse Clariom S Array PSR1400003283.mm.1 NM_013473.4 Refseq
Affymetrix Mouse Clariom S Array PSR1400003283.mm.1 TC1400000368.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR1400003284.mm.1 NM_001281845.1 Refseq
Affymetrix Mouse Clariom S Array PSR1400003284.mm.1 NM_013473.4 Refseq
Affymetrix Mouse Clariom S Array PSR1400003284.mm.1 TC1400000368.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR1400003285.mm.1 NM_001281845.1 Refseq
Affymetrix Mouse Clariom S Array PSR1400003285.mm.1 NM_013473.4 Refseq
Affymetrix Mouse Clariom S Array PSR1400003285.mm.1 TC1400000368.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0