This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm00506502_m1
See other STK38 GE Assays ›
Gene Symbol:
Stk38
Gene Name:
serine/threonine kinase 38
Gene Aliases:
5830476G13Rik, 9530097A09Rik, AA617404, Ndr1
Chromosome Location:
Chr.17: 28970885 - 29008006 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm00506502_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Stk38
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_134115.2 NP_598876.1 10 - 11 1177 83
GenBank mRNA AY292399.1 - 10 - 11 83
BC009658.1 - 10 - 11 1177 3484764 83

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC1700001774.mm.1

Set Membership:

Ambion siRNA available Amplicon length between 71 and 85 Protein Kinase Mammalian Gene Collection Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Stk38

Panther Classification:

Molecular Function -

non-receptor serine/threonine protein kinase

Gene Ontology Categories:

Function(s) Process(es)

cellular protein modification process
protein phosphorylation
phosphorylation
intracellular signal transduction
negative regulation of MAP kinase activity
nucleotide binding
magnesium ion binding
protein kinase activity
protein serine/threonine kinase activity
protein binding
ATP binding
kinase activity
transferase activity
transferase activity, transferring phosphorus-containing groups
mitogen-activated protein kinase kinase kinase binding
metal ion binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR1700016360.mm.1 NM_134115.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR1700016360.mm.1 NM_134115.2 Refseq
Affymetrix Mouse Clariom S Array PSR1700016360.mm.1 TC1700001774.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR1700016358.mm.1 NM_134115.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR1700016358.mm.1 NM_134115.2 Refseq
Affymetrix Mouse Clariom S Array PSR1700016358.mm.1 TC1700001774.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0