This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm00506214_g1
See other DDX1 GE Assays ›
Gene Symbol:
Ddx1
Gene Name:
DEAD (Asp-Glu-Ala-Asp) box polypeptide 1
Gene Aliases:
AA409185, DBP-RB
Chromosome Location:
Chr.12: 13219307 - 13249174 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm00506214_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Ddx1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_134040.1 NP_598801.1 22 - 23 1892 62
GenBank mRNA AK028341.1 - 22 - 23 1889 62
AK132157.1 - 22 - 23 1932 62
AK153335.1 - 22 - 23 1916 62
AK160982.1 - 22 - 23 1893 62
AK166146.1 - 22 - 23 1875 62
BC010624.1 - 22 - 23 1892 3153029 62

Back To Top

More Information


Set Membership:

Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Ddx1

Gene Ontology Categories:

Function(s) Process(es)

double-strand break repair
transcription, DNA-templated
regulation of transcription, DNA-templated
tRNA splicing, via endonucleolytic cleavage and ligation
mRNA processing
tRNA processing
response to virus
RNA secondary structure unwinding
DNA duplex unwinding
response to exogenous dsRNA
nucleic acid phosphodiester bond hydrolysis
regulation of nucleic acid-templated transcription
nucleotide binding
nucleic acid binding
DNA binding
chromatin binding
transcription cofactor activity
RNA binding
double-stranded RNA binding
ATP-dependent RNA helicase activity
helicase activity
nuclease activity
exonuclease activity
protein binding
ATP binding
ATP-dependent helicase activity
poly(A) binding
hydrolase activity
DNA/RNA helicase activity
poly(A) RNA binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-649bfd9c6b-jlsz7:80/100.66.128.142:80.
git-commit: 6001d82975f0049f9efc09ec70b1b2d89229eeb3
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0