This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm00503768_g1
See other CIZ1 GE Assays ›
Gene Symbol:
Ciz1
Gene Name:
CDKN1A interacting zinc finger protein 1
Gene Aliases:
0610038H21Rik, 2900056O04Rik, LSFR1, ZNF356
Chromosome Location:
Chr.2: 32363006 - 32378313 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm00503768_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Ciz1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001252534.1 NP_001239463.1 13 - 14 2197 83
NM_001252536.1 NP_001239465.1 14 - 15 1909 83
NM_001252537.1 NP_001239466.1 13 - 14 2170 83
NM_001252538.1 NP_001239467.1 13 - 14 1860 83
NM_028412.2 NP_082688.1 12 - 13 1974 83
GenBank mRNA AJ575057.1 - 11 - 12 1481 83
AK089986.1 - 13 - 14 2173 83
AK137961.1 - 13 - 14 1776 83
AK159103.1 - 13 - 14 2175 83
AK165687.1 - 14 - 15 1863 83
BC018483.1 - 12 - 13 1938 3597692 83
BC072624.1 - 3060 6825309 83

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC0200000577.mm.1

Set Membership:

Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Ciz1

Gene Ontology Categories:

Function(s) Process(es)

positive regulation of DNA-dependent DNA replication initiation
maintenance of protein location in nucleus
cyclin binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0200005190.mm.1 TC0200000577.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0200005193.mm.1 NM_001252534.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0200005193.mm.1 NM_001252534.1 Refseq
Affymetrix Mouse Clariom S Array PSR0200005193.mm.1 NM_001252536.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0200005193.mm.1 NM_001252536.1 Refseq
Affymetrix Mouse Clariom S Array PSR0200005193.mm.1 NM_001252537.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0200005193.mm.1 NM_001252537.1 Refseq
Affymetrix Mouse Clariom S Array PSR0200005193.mm.1 NM_001252538.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0200005193.mm.1 NM_001252538.1 Refseq
Affymetrix Mouse Clariom S Array PSR0200005193.mm.1 NM_028412.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR0200005193.mm.1 NM_028412.2 Refseq
Affymetrix Mouse Clariom S Array PSR0200005193.mm.1 TC0200000577.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0200005195.mm.1 NM_001252534.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0200005195.mm.1 NM_001252534.1 Refseq
Affymetrix Mouse Clariom S Array PSR0200005195.mm.1 NM_001252536.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0200005195.mm.1 NM_001252536.1 Refseq
Affymetrix Mouse Clariom S Array PSR0200005195.mm.1 NM_001252537.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0200005195.mm.1 NM_001252537.1 Refseq
Affymetrix Mouse Clariom S Array PSR0200005195.mm.1 NM_001252538.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0200005195.mm.1 NM_001252538.1 Refseq
Affymetrix Mouse Clariom S Array PSR0200005195.mm.1 NM_028412.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR0200005195.mm.1 NM_028412.2 Refseq
Affymetrix Mouse Clariom S Array PSR0200005195.mm.1 TC0200000577.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0200005189.mm.1 TC0200000577.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-649bfd9c6b-jlsz7:80/100.66.128.142:80.
git-commit: 6001d82975f0049f9efc09ec70b1b2d89229eeb3
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0