This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm00503218_g1
See other CUTA GE Assays ›
Gene Symbol:
Cuta
Gene Name:
cutA divalent cation tolerance homolog (E. coli)
Gene Aliases:
0610039D01Rik, 1810022E02Rik, 1810060C03Rik, 2700094G22Rik, AI326454
Chromosome Location:
Chr.17: 26937972 - 26939815 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm00503218_g1
Size
Availability Made To Order
Catalog # 4351372
Price 4,408.00
Your Price
Online offer:
4408.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Cuta
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_026307.3 NP_080583.3 5 - 6 461 98
NM_026948.3 NP_081224.1 4 - 5 409 98
NR_104458.1 4 - 5 495 130
GenBank mRNA AK002828.1 - 5 - 6 405 98
AK003381.1 - 4 - 5 349 98
AK003647.1 - 5 - 6 403 98
AK004197.1 - 5 - 6 403 98
AK007580.1 - 3 - 4 538 207
AK007912.1 - 4 - 5 484 130
AK145185.1 - 563 207
AK159336.1 - 5 - 6 408 98
BC021463.1 - 3 - 4 523 98
BC024422.1 - 5 - 6 391 5005082 98
BC028531.1 - 4 - 5 500 207
CA323205.1 - 4 - 5 375 98
CK624897.1 - 4 - 5 386 98

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC1700001711.mm.1

Set Membership:

Ambion siRNA available Amplicon length between 86 and 100 Amplicon length greater than or equal to 101 Transporter Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Cuta

Panther Classification:

Molecular Function -

primary active transporter

Gene Ontology Categories:

Function(s) Process(es)

protein localization
response to metal ion
protein homooligomerization
enzyme binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR1700015826.mm.1 NM_026307.3 Refseq
Affymetrix Mouse Clariom S Array PSR1700015826.mm.1 NM_026948.3 Refseq
Affymetrix Mouse Clariom S Array PSR1700015826.mm.1 NR_104458.1 Refseq
Affymetrix Mouse Clariom S Array PSR1700015826.mm.1 TC1700001711.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-649bfd9c6b-jlsz7:80/100.66.128.142:80.
git-commit: 6001d82975f0049f9efc09ec70b1b2d89229eeb3
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0