This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm00502960_m1
See other THEM5 GE Assays ›
Gene Symbol:
Them5
Gene Name:
thioesterase superfamily member 5
Gene Aliases:
1110007B02Rik, 1110038F21Rik
Chromosome Location:
Chr.3: 94342099 - 94347352 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm00502960_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Them5
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_025416.3 NP_079692.1 1 - 2 283 115
GenBank mRNA AK004155.1 - 1 - 2 284 115
AK009228.1 - 1 - 2 282 115
BC046437.1 - 1 - 2 265 3969280 115

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC0300000862.mm.1

Set Membership:

Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Them5

Panther Classification:

Molecular Function -

esterase hydrolase metabolite interconversion enzyme

Gene Ontology Categories:

Function(s) Process(es)

lipid metabolic process
fatty acid metabolic process
long-chain fatty-acyl-CoA metabolic process
cardiolipin acyl-chain remodeling
palmitoyl-CoA hydrolase activity
hydrolase activity

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0300006575.mm.1 NM_025416.3.1 Refseq
Affymetrix Mouse Clariom S Array PSR0300006575.mm.1 NM_025416.3 Refseq
Affymetrix Mouse Clariom S Array PSR0300006575.mm.1 TC0300000862.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0300006576.mm.1 NM_025416.3.1 Refseq
Affymetrix Mouse Clariom S Array PSR0300006576.mm.1 NM_025416.3 Refseq
Affymetrix Mouse Clariom S Array PSR0300006576.mm.1 TC0300000862.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-69fc894774-tvlwx:80/100.66.128.142:80.
git-commit: 8e82defa9bf25b3b77800b5e306013d4ba941391
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0