This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm00502905_m1
See other FOPNL GE Assays ›
Gene Symbol:
Fopnl
Gene Name:
Fgfr1op N-terminal like
Gene Aliases:
0610037P05Rik
Chromosome Location:
Chr.16: 14299241 - 14317926 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm00502905_m1
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Fopnl
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_025345.2 NP_079621.1 4 - 5 463 75
GenBank mRNA AK002785.1 - 2 - 3 193 75
AK012206.1 - 4 - 5 458 75
AK146884.1 - 4 - 5 489 75
AK153894.1 - 4 - 5 467 75
BC011300.1 - 4 - 5 463 3671642 75

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC1600001248.mm.1

Set Membership:

Ambion siRNA available Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Mus musculus 1700 Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Fopnl

Gene Ontology Categories:

Function(s) Process(es)

cell projection organization
microtubule anchoring
cilium assembly
molecular_function

MicroArray Probes:

Agilent Affymetrix ABI Illumina

Array Name Probe IDs Targets Target Type
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P30561658 - -
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P30561659 - -
Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1955791 BC011300.1 mouse_transcriptome
Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1955791 AK146884.1 mouse_transcriptome
Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1955791 NM_025345.2 mouse_transcriptome
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 847307 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 87918 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 278708 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 276778 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 409537 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 637230 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 23473 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 251470 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 539313 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 363277 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 557385 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 734172 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 255511 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 654399 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 915630 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 89468 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 201241 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 948244 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 133943 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 20117 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 323093 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 174093 - -
Agilent SurePrint G3 Mouse CGH Microarray 4x180K 86875 - -
Agilent SurePrint G3 Mouse CGH Microarray 4x180K 140429 - -
Agilent SurePrint G3 Mouse CGH Microarray 4x180K 132990 - -
Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P108509 - -
Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P147442 - -
Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P120632 - -
Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P106671 - -
Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P179732 - -
Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P158481 - -
Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P152427 - -
Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P109518 - -
SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1955791 BC011300.1 mouse_transcriptome
SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1955791 AK146884.1 mouse_transcriptome
SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1955791 NM_025345.2 mouse_transcriptome
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30561652 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30561645 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30561671 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30561650 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30561649 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30561667 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30561670 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30561663 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30561640 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30561676 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30561642 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30561673 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30561674 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30561661 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30561656 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30561644 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30561659 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30561668 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30561647 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30561657 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30561658 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30561654 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30561653 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30561672 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30561664 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30561678 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30561675 - -
Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P08514596 - -
Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P08514574 - -
Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P03136325 - -
Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P03136574 - -
Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P03136306 - -
Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P03136337 - -
Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P152427 - -
Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P120632 - -
Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P08514468 - -
Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P08514640 - -
Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P03136320 - -
Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P158481 - -
Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P08515036 - -
Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P08515026 - -
Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P109518 - -
Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P147442 - -
Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P03136317 - -
Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P03136269 - -
Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P108509 - -
Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P08514440 - -
Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P179732 - -
Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P106671 - -
Whole Mouse Genome G4122A A_51_P125096 BC011300.1 mouse_transcriptome
Whole Mouse Genome G4122A A_51_P125096 AK146884.1 mouse_transcriptome
Whole Mouse Genome G4122A A_51_P125096 NM_025345.2 mouse_transcriptome
Whole Mouse Genome G4122A A_52_P236994 - -
Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P102755 - -
Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P147442 - -
Agilent Mouse Genome CGH Microarray 105A (G4416A) A_67_P03136306 - -
Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P109518 - -
Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P108509 - -
Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P152427 - -
Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P179732 - -
Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P106671 - -
Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P158481 - -
Agilent Mouse Genome CGH Microarray 105A (G4416A) A_67_P03136320 - -
Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P120632 - -
Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P12789862 - -
Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P12789868 - -
Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P12892362 - -
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P30561658 - -
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P30561659 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30561537 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30561649 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30561586 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30561588 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30561495 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30561642 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30561501 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30561562 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30561608 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30561675 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30561565 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30561601 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30561573 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30561671 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30561541 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30561579 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30561521 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30561654 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30561511 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30561635 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30561606 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30561484 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30561506 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30561497 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30561566 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30561583 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30561544 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30561523 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30561591 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30561571 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30561567 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30561623 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30561490 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30561621 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30561597 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30561552 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30561663 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30561595 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30561570 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30561633 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30561517 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30561507 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30561575 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30561584 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30561603 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30561636 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30561496 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30561533 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30561492 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30561672 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30561647 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30561508 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30561600 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30561529 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30561561 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30561576 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30561665 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30561614 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30561652 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30561644 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30561640 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30561659 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30561500 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30561514 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30561667 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30561513 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30561650 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30561656 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30561488 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30561527 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30561482 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30561548 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30561673 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30561610 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30561509 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30561657 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30561551 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30561520 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30561535 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30561542 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30561532 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30561577 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30561629 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30561618 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30561485 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30561569 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30561505 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30561668 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30561502 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30561604 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30561653 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30561615 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30561493 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P30561678 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P30561645 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P13035032 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P30561665 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P30561652 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P30561644 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P30561642 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P30561650 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P30561629 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P30561653 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P30561627 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P30561625 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P30561632 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P30561664 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P12789838 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P30561635 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P30561614 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P30561617 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P30561675 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P12892362 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P12892363 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P30561672 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P12787765 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P30561649 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P12637380 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P30561667 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P30561545 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P30561547 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P30561639 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P30561647 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P30561621 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P30561659 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P30561631 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P12789866 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P30561673 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P30561623 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P30561668 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P30561633 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P30561655 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P30561671 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P30561638 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P30561640 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P30561615 - -
Agilent Six_ChIP_03_of_03_v2 A_68_P30561636 - -
Array Name Probe IDs Targets Target Type
Affymetrix Mouse Gene 1.0 ST Array 10437932 - -
Affymetrix Mouse Gene 1.0 ST Array 10437929 - -
Affymetrix Mouse Gene 1.0 ST Array 10437930 - -
Affymetrix Mouse Gene 1.0 ST Array 10437933 - -
Affymetrix Mouse Gene 1.0 ST Array 10437931 - -
Affymetrix HT MG-430 PM Array Plate 1451187_PM_at - -
Affymetrix Mouse Gene 1.1 ST Array 10437928 - -
Mouse Genome 430 2.0 Array 1451187_PM_at - -
Mouse Genome 430 2.0 Array 1451187_at - -
Affymetrix GeneChip Mouse Genome 430 2.0 Array 66086_PM_at - -
Affymetrix GeneChip Mouse Genome 430 2.0 Array 66086_at - -
Affymetrix Mouse Clariom S Array PSR1600010047.mm.1 NM_025345.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR1600010047.mm.1 NM_025345.2 Refseq
Affymetrix Mouse Clariom S Array PSR1600010047.mm.1 TC1600001248.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR1600010048.mm.1 NM_025345.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR1600010048.mm.1 NM_025345.2 Refseq
Affymetrix Mouse Clariom S Array PSR1600010048.mm.1 TC1600001248.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR1600010046.mm.1 NM_025345.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR1600010046.mm.1 NM_025345.2 Refseq
Affymetrix Mouse Clariom S Array PSR1600010046.mm.1 TC1600001248.mm.1 MTA TC
Mouse Expression Set 430 1451187_PM_at - -
Mouse Expression Set 430 1451187_at - -
Murine Genome U74v2 Set 165469_PM_f_at - -
Murine Genome U74v2 Set 165469_f_at - -
Mouse Genome 430A 2.0 Array 1451187_at - -
Mouse Genome 430A 2.0 Array 1451187_PM_at - -
Affymetrix HT Mouse Genome 430A Array 1451187_PM_at - -
Affymetrix HT Mouse Genome 430A Array 1451187_at - -
Array Name Probe IDs Targets Target Type
ABI Mouse Genome Survey Microarray 779097 - -
Array Name Probe IDs Targets Target Type
Illumina mouseRef-8 v1.1 expression beadchip ILMN_1230077 - -
Illumina mouseRef-8 v1.1 expression beadchip ILMN_2614091 - -
Illumina mouseRef-8 v1.1 expression beadchip ILMN_2717539 - -
Illumina mouse-6 v1.1 expression beadchip ILMN_2614091 - -
Illumina mouse-6 v1.1 expression beadchip ILMN_2717539 - -
Illumina mouse-6 v1.1 expression beadchip ILMN_1230077 - -
MouseRef-8 v2 Expression BeadChip ILMN_2717539 - -

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0