This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm00501968_m1
See other GNB4 GE Assays ›
Gene Symbol:
Gnb4
Gene Name:
guanine nucleotide binding protein (G protein), beta 4
Gene Aliases:
6720453A21Rik, G(beta)4
Chromosome Location:
Chr.3: 32580332 - 32616535 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm00501968_m1
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Gnb4
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_013531.4 NP_038559.2 4 - 5 340 68
GenBank mRNA AF277893.1 - 3 - 4 204 68
AF501886.1 - 3 - 4 204 68
AK036816.1 - 4 - 5 342 68
AK098139.1 - 4 - 5 273 68
AK152227.1 - 4 - 5 331 68
AK167416.1 - 4 - 5 373 68
AK168302.1 - 4 - 5 296 68
AK169752.1 - 4 - 5 331 68
AK172147.1 - 5 - 6 473 68
AK187079.1 - 1 - 2 104 68
BC028753.1 - 4 - 5 311 1348753 68
M63658.1 - 3 - 4 244 68
M87286.1 - 3 - 4 244 68
S86124.1 - 3 - 4 244 68

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC0300001802.mm.1

Set Membership:

Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Gnb4

Panther Classification:

Molecular Function -

heterotrimeric G-protein

Gene Ontology Categories:

Function(s) Process(es)

signal transduction
G-protein coupled receptor signaling pathway
GTPase activity
signal transducer activity
protein complex binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0300013960.mm.1 NM_013531.5 Refseq
Affymetrix Mouse Clariom S Array PSR0300013960.mm.1 TC0300001802.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0