This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm00499505_m1
See other LIMS1 GE Assays ›
Gene Symbol:
Lims1
Gene Name:
LIM and senescent cell antigen-like domains 1
Gene Aliases:
2310016J22Rik, 4921524A02Rik, AI507642, AU021743, AW551584, C430041B13Rik, Lims1l, PINCH, PINCH1
Chromosome Location:
Chr.10: 58323435 - 58424691 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm00499505_m1
Size
Availability Made To Order
Catalog # 4351372
Price 4,408.00
Your Price
Online offer:
4408.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Lims1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001193303.1 NP_001180232.1 6 - 7 875 91
NM_026148.3 NP_080424.2 6 - 7 869 91
NM_201242.1 NP_957694.1 6 - 7 893 91
GenBank mRNA AK009390.1 - 6 - 7 876 91
AK014954.1 - 6 - 7 870 91
AK082932.1 - 6 - 7 738 91
AK088939.1 - 6 - 7 744 91
AK151235.1 - 6 - 7 744 91
AK151938.1 - 6 - 7 736 91
AK152679.1 - 6 - 7 736 91
AK153346.1 - 6 - 7 744 91
BC005621.1 - 6 - 7 724 3711611 91
BC067393.1 - 6 - 7 667 91
DQ003608.1 - 6 - 7 860 91
EU122953.1 - 6 - 7 859 91
EU122954.1 - 6 - 7 727 91

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC1000000514.mm.1

Set Membership:

Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Lims1

Panther Classification:

Molecular Function -

cell junction protein

Gene Ontology Categories:

Function(s) Process(es)

cell-matrix adhesion
establishment or maintenance of cell polarity
positive regulation of gene expression
positive regulation of cell-substrate adhesion
single organismal cell-cell adhesion
chordate embryonic development
negative regulation of apoptotic process
positive regulation of GTPase activity
establishment of protein localization
cell-cell junction organization
negative regulation of transcription, DNA-templated
negative regulation of epithelial cell proliferation
protein heterooligomerization
positive regulation of focal adhesion assembly
positive regulation of substrate adhesion-dependent cell spreading
negative regulation of neural precursor cell proliferation
negative regulation of hepatocyte proliferation
positive regulation of integrin-mediated signaling pathway
protein binding
zinc ion binding
protein kinase binding
metal ion binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR1000003412.mm.1 NM_001193303.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR1000003412.mm.1 NM_001193303.1 Refseq
Affymetrix Mouse Clariom S Array PSR1000003412.mm.1 NM_026148.3.1 Refseq
Affymetrix Mouse Clariom S Array PSR1000003412.mm.1 NM_026148.3 Refseq
Affymetrix Mouse Clariom S Array PSR1000003412.mm.1 NM_201242.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR1000003412.mm.1 NM_201242.1 Refseq
Affymetrix Mouse Clariom S Array PSR1000003412.mm.1 TC1000000514.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-649bfd9c6b-jlsz7:80/100.66.128.142:80.
git-commit: 6001d82975f0049f9efc09ec70b1b2d89229eeb3
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0